The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001251977.1 | NP_001238906.1 | 4 - 5 | 815 | 88 |
NM_001251978.1 | NP_001238907.1 | 4 - 5 | 729 | 88 | |
NM_001251979.1 | NP_001238908.1 | 4 - 5 | 707 | 88 | |
NM_001251980.1 | NP_001238909.1 | 3 - 4 | 564 | 88 | |
NM_001251981.1 | NP_001238910.1 | 3 - 4 | 641 | 88 | |
NM_001251983.1 | NP_001238912.1 | 3 - 4 | 594 | 88 | |
NM_001251984.1 | NP_001238913.1 | 3 - 4 | 453 | 88 | |
NM_013441.3 | NP_038469.1 | 4 - 5 | 848 | 88 | |
GenBank mRNA | AF176116.1 | - | 3 - 4 | 557 | 88 |
AF176117.1 | - | 3 - 4 | 449 | 88 | |
BC035854.1 | - | 4 - 5 | 848 | 88 | |
BX105019.1 | - | 2 - 3 | 275 | 88 | |
CT001954.1 | - | 3 - 4 | 588 | 88 | |
EF431960.1 | - | 2 - 3 | 387 | 88 | |
EF529733.1 | - | 2 - 3 | 387 | 88 | |
HQ287726.1 | - | 3 - 4 | 377 | 88 | |
HQ317421.1 | - | 4 - 5 | 683 | 88 | |
HQ317422.1 | - | 3 - 4 | 429 | 88 | |
HQ317423.1 | - | 4 - 5 | 729 | 88 | |
HQ317426.1 | - | 3 - 4 | 598 | 88 | |
HQ317427.1 | - | 2 - 3 | 344 | 88 | |
JN203053.1 | - | 2 - 3 | 424 | 88 |