The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014003.3 | NP_054722.2 | 1 - 2 | 331 | 117 |
XM_005256269.1 | XP_005256326.1 | 1 - 2 | 258 | 117 | |
XM_011523484.1 | XP_011521786.1 | 1 - 2 | 258 | 117 | |
XM_017023913.1 | XP_016879402.1 | 1 - 2 | 258 | 117 | |
GenBank mRNA | AK291634.1 | - | 1 - 2 | 123 | 117 |
AK293776.1 | - | 1 - 2 | 123 | 117 | |
AK297269.1 | - | 1 - 2 | 123 | 117 | |
AK301074.1 | - | 1 - 2 | 200 | 117 | |
AK311169.1 | - | 1 - 2 | 120 | 117 | |
BC004235.2 | - | 1 - 2 | 107 | 117 | |
BC008340.1 | - | 1 - 2 | 107 | 117 | |
BG719187.1 | - | 1 - 2 | 338 | 117 | |
BM473438.1 | - | 1 - 2 | 231 | 117 | |
D86977.1 | - | 1 - 2 | 112 | 117 |