Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX24
Entrez Gene ID
28966
Gene Name
sorting nexin 24
Gene Aliases
PRO1284, SBBI31
Chromosome Location
Chr.5: 122845465 - 123029318 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014035.2 NP_054754.1 1 - 2 225 120
XM_005271972.4 XP_005272029.1 1 - 2 114 120
XM_006714592.3 XP_006714655.1 1 - 2 180 120
GenBank mRNA AF139461.1 - 1 - 2 114 120
AY044655.1 - 1 - 2 56 120
AY358098.1 - 1 - 2 74 120
BC010886.1 - 1 - 2 185 120
BC069012.1 - 1 - 2 172 120
BQ278046.1 - 1 - 2 197 120
DB498436.1 - 1 - 2 241 120

View Full Product Details