The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159767.1 | NP_001153239.1 | 3 - 4 | 417 | 78 |
NM_014038.2 | NP_054757.1 | 3 - 4 | 401 | 78 | |
NR_027624.1 | 3 - 4 | 417 | 78 | ||
XM_006715706.1 | XP_006715769.1 | 3 - 4 | 285 | 78 | |
XM_006715707.1 | XP_006715770.1 | 3 - 4 | 368 | 78 | |
XM_006715708.1 | XP_006715771.1 | 3 - 4 | 287 | 78 | |
GenBank mRNA | AF083246.1 | - | 3 - 4 | 401 | 78 |
AF110323.1 | - | 2 - 3 | 239 | 78 | |
AK001218.1 | - | 3 - 4 | 381 | 78 | |
AK001260.1 | - | 3 - 4 | 375 | 78 | |
AK027837.1 | - | 3 - 4 | 285 | 78 | |
AK300655.1 | - | 3 - 4 | 346 | 78 | |
BC003056.1 | - | 3 - 4 | 327 | 78 | |
BC008453.1 | - | 3 - 4 | 354 | 78 | |
BC009597.1 | - | 3 - 4 | 385 | 78 | |
BC017794.1 | - | 3 - 4 | 272 | 78 | |
BC107757.1 | - | 3 - 4 | 375 | 78 |