The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316957.1 | NP_001303886.1 | 3 - 4 | 322 | 82 |
NM_001316958.1 | NP_001303887.1 | 3 - 4 | 322 | 82 | |
NM_001316960.1 | NP_001303889.1 | 3 - 4 | 398 | 82 | |
NM_001316961.1 | NP_001303890.1 | 3 - 4 | 318 | 82 | |
NM_001316962.1 | NP_001303891.1 | 3 - 4 | 322 | 82 | |
NM_024684.3 | NP_078960.1 | 2 - 3 | 280 | 82 | |
NR_133637.1 | 2 - 3 | 280 | 82 | ||
XM_011544968.2 | XP_011543270.1 | 2 - 3 | 280 | 82 | |
XM_011544969.2 | XP_011543271.1 | 2 - 3 | 273 | 82 | |
XM_017017620.1 | XP_016873109.1 | 2 - 3 | 302 | 82 | |
GenBank mRNA | AF092136.1 | - | 3 - 4 | 281 | 82 |
AI569731.1 | - | 2 - 3 | 82 | ||
AK024688.1 | - | 2 - 3 | 239 | 82 | |
BC002752.2 | - | 2 - 3 | 232 | 82 | |
BC016854.2 | - | 2 - 3 | 209 | 82 | |
BI791786.1 | - | 3 - 4 | 331 | 82 | |
BM930331.1 | - | 3 - 4 | 267 | 82 | |
BP270361.1 | - | 2 - 3 | 280 | 82 | |
BX646013.1 | - | 2 - 3 | 264 | 82 | |
CN362204.1 | - | 3 - 4 | 293 | 82 | |
CR457332.1 | - | 1 - 2 | 134 | 82 |