Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202036233-202036731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD7
Entrez Gene ID
8324
Gene Name
frizzled class receptor 7
Gene Aliases
FzE3
UniGene
Hs.173859
Gene Chromosome Location
Chr.2: 202034587-202038437 on Build GRCh38
Transcripts
NM_003507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6156564 c.1687A>T p.R563* Chr.2: 202036334-202036334 on Build GRCh38
4832294 c.1680C>G p.F560L Chr.2: 202036327-202036327 on Build GRCh38
1736252 c.1718C>T p.A573V Chr.2: 202036365-202036365 on Build GRCh38
1404559 c.1672C>T p.R558C Chr.2: 202036319-202036319 on Build GRCh38
6437615 c.1693A>T p.S565C Chr.2: 202036340-202036340 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs185785726 Chr.2: 202036619-202036619 on Build GRCh38
rs758853443 Chr.2: 202036276-202036276 on Build GRCh38
rs769424376 Chr.2: 202036288-202036288 on Build GRCh38
rs748151902 Chr.2: 202036403-202036403 on Build GRCh38
rs754937674 Chr.2: 202036416-202036418 on Build GRCh38
rs932729482 Chr.2: 202036478-202036478 on Build GRCh38
rs1015641916 Chr.2: 202036549-202036549 on Build GRCh38
rs753302518 Chr.2: 202036366-202036366 on Build GRCh38
rs761575550 Chr.2: 202036322-202036322 on Build GRCh38
rs771841493 Chr.2: 202036404-202036404 on Build GRCh38
rs546449437 Chr.2: 202036377-202036377 on Build GRCh38
rs534588552 Chr.2: 202036407-202036407 on Build GRCh38
rs755768740 Chr.2: 202036380-202036380 on Build GRCh38
rs992370051 Chr.2: 202036651-202036651 on Build GRCh38
rs940058855 Chr.2: 202036560-202036560 on Build GRCh38
rs900336952 Chr.2: 202036468-202036468 on Build GRCh38
rs556682682 Chr.2: 202036696-202036696 on Build GRCh38
rs1025128012 Chr.2: 202036655-202036655 on Build GRCh38
rs199589707 Chr.2: 202036389-202036389 on Build GRCh38
rs754291190 Chr.2: 202036265-202036265 on Build GRCh38
rs1033879510 Chr.2: 202036374-202036374 on Build GRCh38
rs773419766 Chr.2: 202036336-202036336 on Build GRCh38
rs371345546 Chr.2: 202036382-202036382 on Build GRCh38
rs977722364 Chr.2: 202036672-202036672 on Build GRCh38
rs774179533 Chr.2: 202036303-202036303 on Build GRCh38
rs755458386 Chr.2: 202036267-202036267 on Build GRCh38
rs774554658 Chr.2: 202036552-202036552 on Build GRCh38
rs538424242 Chr.2: 202036658-202036658 on Build GRCh38
rs766517331 Chr.2: 202036260-202036260 on Build GRCh38
rs755782157 Chr.2: 202036695-202036695 on Build GRCh38
rs111263028 Chr.2: 202036273-202036273 on Build GRCh38
rs995764163 Chr.2: 202036606-202036606 on Build GRCh38
rs768569299 Chr.2: 202036296-202036296 on Build GRCh38
rs977753392 Chr.2: 202036419-202036419 on Build GRCh38
rs772119967 Chr.2: 202036584-202036584 on Build GRCh38
rs955763827 Chr.2: 202036456-202036456 on Build GRCh38
rs13034206 Chr.2: 202036512-202036512 on Build GRCh38
rs752391872 Chr.2: 202036665-202036665 on Build GRCh38
rs547551444 Chr.2: 202036428-202036428 on Build GRCh38
rs974234940 Chr.2: 202036367-202036367 on Build GRCh38
rs765723806 Chr.2: 202036360-202036360 on Build GRCh38
rs1038809392 Chr.2: 202036452-202036452 on Build GRCh38
rs759636568 Chr.2: 202036416-202036416 on Build GRCh38
rs536312561 Chr.2: 202036483-202036483 on Build GRCh38
rs528874438 Chr.2: 202036281-202036281 on Build GRCh38
rs759567940 Chr.2: 202036354-202036354 on Build GRCh38
rs1038473049 Chr.2: 202036574-202036574 on Build GRCh38
rs925202604 Chr.2: 202036424-202036424 on Build GRCh38
rs1044691812 Chr.2: 202036646-202036646 on Build GRCh38
rs772937162 Chr.2: 202036406-202036406 on Build GRCh38
rs559288287 Chr.2: 202036617-202036617 on Build GRCh38
rs989868994 Chr.2: 202036479-202036479 on Build GRCh38
rs745552067 Chr.2: 202036282-202036282 on Build GRCh38
rs748503156 Chr.2: 202036275-202036275 on Build GRCh38
rs748759824 Chr.2: 202036290-202036290 on Build GRCh38
rs948781606 Chr.2: 202036631-202036631 on Build GRCh38
rs574540529 Chr.2: 202036677-202036678 on Build GRCh38
rs368139563 Chr.2: 202036263-202036263 on Build GRCh38
rs750086736 Chr.2: 202036378-202036378 on Build GRCh38
rs534534598 Chr.2: 202036566-202036566 on Build GRCh38
rs372045596 Chr.2: 202036289-202036289 on Build GRCh38
rs921567393 Chr.2: 202036522-202036522 on Build GRCh38
rs959823653 Chr.2: 202036644-202036644 on Build GRCh38
rs934142517 Chr.2: 202036528-202036528 on Build GRCh38
rs1015839962 Chr.2: 202036337-202036337 on Build GRCh38
rs987013102 Chr.2: 202036536-202036536 on Build GRCh38
rs771894898 Chr.2: 202036332-202036332 on Build GRCh38
rs565714072 Chr.2: 202036340-202036340 on Build GRCh38
rs891636768 Chr.2: 202036682-202036682 on Build GRCh38
rs766526311 Chr.2: 202036342-202036342 on Build GRCh38
rs112052844 Chr.2: 202036422-202036422 on Build GRCh38
rs13034579 Chr.2: 202036556-202036556 on Build GRCh38
rs891238193 Chr.2: 202036493-202036493 on Build GRCh38
rs574857420 Chr.2: 202036705-202036705 on Build GRCh38
rs779576113 Chr.2: 202036384-202036384 on Build GRCh38
rs367875554 Chr.2: 202036410-202036410 on Build GRCh38
rs373957083 Chr.2: 202036386-202036386 on Build GRCh38
rs758800464 Chr.2: 202036375-202036375 on Build GRCh38
rs771147451 Chr.2: 202036408-202036408 on Build GRCh38

View Full Product Details