Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202036632-202037140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD7
Entrez Gene ID
8324
Gene Name
frizzled class receptor 7
Gene Aliases
FzE3
UniGene
Hs.173859
Gene Chromosome Location
Chr.2: 202034587-202038437 on Build GRCh38
Transcripts
NM_003507.1

SNP Detail

SNP ID Chromosome Location
rs542261541 Chr.2: 202036872-202036872 on Build GRCh38
rs1014170360 Chr.2: 202036903-202036903 on Build GRCh38
rs900274011 Chr.2: 202037073-202037073 on Build GRCh38
rs560295211 Chr.2: 202037011-202037011 on Build GRCh38
rs750687130 Chr.2: 202036860-202036860 on Build GRCh38
rs966399117 Chr.2: 202036959-202036959 on Build GRCh38
rs891627025 Chr.2: 202036956-202036956 on Build GRCh38
rs572100047 Chr.2: 202037025-202037025 on Build GRCh38
rs1042775432 Chr.2: 202037068-202037068 on Build GRCh38
rs955626618 Chr.2: 202036786-202036786 on Build GRCh38
rs996347323 Chr.2: 202037114-202037114 on Build GRCh38
rs891202300 Chr.2: 202036784-202036784 on Build GRCh38
rs556682682 Chr.2: 202036696-202036696 on Build GRCh38
rs1025128012 Chr.2: 202036655-202036655 on Build GRCh38
rs940120987 Chr.2: 202036794-202036794 on Build GRCh38
rs977722364 Chr.2: 202036672-202036672 on Build GRCh38
rs564817033 Chr.2: 202037056-202037056 on Build GRCh38
rs933135278 Chr.2: 202036746-202036746 on Build GRCh38
rs1028977393 Chr.2: 202036757-202036757 on Build GRCh38
rs1031945178 Chr.2: 202036972-202036972 on Build GRCh38
rs898500108 Chr.2: 202036839-202036839 on Build GRCh38
rs867081764 Chr.2: 202036951-202036951 on Build GRCh38
rs538424242 Chr.2: 202036658-202036658 on Build GRCh38
rs987472542 Chr.2: 202036838-202036838 on Build GRCh38
rs755782157 Chr.2: 202036695-202036695 on Build GRCh38
rs921824236 Chr.2: 202037036-202037036 on Build GRCh38
rs780493290 Chr.2: 202037048-202037048 on Build GRCh38
rs974651011 Chr.2: 202036730-202036730 on Build GRCh38
rs752391872 Chr.2: 202036665-202036665 on Build GRCh38
rs974465925 Chr.2: 202036750-202036750 on Build GRCh38
rs554388110 Chr.2: 202036715-202036715 on Build GRCh38
rs928640493 Chr.2: 202036890-202036890 on Build GRCh38
rs1025177381 Chr.2: 202036926-202036926 on Build GRCh38
rs957714226 Chr.2: 202036984-202036984 on Build GRCh38
rs920035316 Chr.2: 202036873-202036873 on Build GRCh38
rs574540529 Chr.2: 202036677-202036678 on Build GRCh38
rs954686963 Chr.2: 202037059-202037059 on Build GRCh38
rs974146273 Chr.2: 202036869-202036869 on Build GRCh38
rs758825582 Chr.2: 202036954-202036954 on Build GRCh38
rs540184112 Chr.2: 202037035-202037035 on Build GRCh38
rs879716668 Chr.2: 202036845-202036845 on Build GRCh38
rs777495723 Chr.2: 202036822-202036822 on Build GRCh38
rs895424679 Chr.2: 202036874-202036874 on Build GRCh38
rs999233818 Chr.2: 202036967-202036967 on Build GRCh38
rs1036997852 Chr.2: 202036800-202036800 on Build GRCh38
rs921759609 Chr.2: 202036736-202036736 on Build GRCh38
rs891636768 Chr.2: 202036682-202036682 on Build GRCh38
rs995733213 Chr.2: 202036855-202036855 on Build GRCh38
rs1044377426 Chr.2: 202036881-202036881 on Build GRCh38
rs745613368 Chr.2: 202036719-202036719 on Build GRCh38
rs574857420 Chr.2: 202036705-202036705 on Build GRCh38
rs761726560 Chr.2: 202036893-202036894 on Build GRCh38
rs974407871 Chr.2: 202037003-202037003 on Build GRCh38
rs1051627057 Chr.2: 202036765-202036765 on Build GRCh38

View Full Product Details