Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202772976-202773496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICA1L
Entrez Gene ID
130026
Gene Name
islet cell autoantigen 1 like
Gene Aliases
ALS2CR14, ALS2CR15
UniGene
Hs.516629
Gene Chromosome Location
Chr.2: 202773150-202871985 on Build GRCh38
Transcripts
NM_001288622.1 NM_001288623.1 NM_138468.5

SNP Detail

SNP ID Chromosome Location
rs368859317 Chr.2: 202773133-202773133 on Build GRCh38
rs779330356 Chr.2: 202773108-202773108 on Build GRCh38
rs374256778 Chr.2: 202773437-202773437 on Build GRCh38
rs1045268043 Chr.2: 202773111-202773111 on Build GRCh38
rs1036120224 Chr.2: 202773034-202773034 on Build GRCh38
rs1024652363 Chr.2: 202773385-202773385 on Build GRCh38
rs7598328 Chr.2: 202773460-202773460 on Build GRCh38
rs943913010 Chr.2: 202773150-202773150 on Build GRCh38
rs555371876 Chr.2: 202773317-202773317 on Build GRCh38
rs769265163 Chr.2: 202773087-202773088 on Build GRCh38
rs572537026 Chr.2: 202773413-202773413 on Build GRCh38
rs977687859 Chr.2: 202773398-202773401 on Build GRCh38
rs80181705 Chr.2: 202773289-202773289 on Build GRCh38
rs180826765 Chr.2: 202773210-202773210 on Build GRCh38
rs536980047 Chr.2: 202773273-202773273 on Build GRCh38
rs7597959 Chr.2: 202773150-202773150 on Build GRCh38
rs1034636643 Chr.2: 202773327-202773327 on Build GRCh38
rs1014395157 Chr.2: 202773378-202773378 on Build GRCh38
rs866261028 Chr.2: 202773142-202773142 on Build GRCh38
rs966440059 Chr.2: 202773394-202773394 on Build GRCh38
rs189445736 Chr.2: 202773149-202773149 on Build GRCh38
rs769911605 Chr.2: 202773053-202773053 on Build GRCh38
rs375079080 Chr.2: 202773288-202773288 on Build GRCh38
rs773425664 Chr.2: 202773081-202773081 on Build GRCh38
rs888496345 Chr.2: 202773307-202773307 on Build GRCh38
rs574817854 Chr.2: 202773322-202773322 on Build GRCh38
rs553839516 Chr.2: 202773064-202773064 on Build GRCh38
rs56253233 Chr.2: 202773212-202773212 on Build GRCh38
rs560657167 Chr.2: 202773401-202773401 on Build GRCh38
rs565591390 Chr.2: 202773099-202773099 on Build GRCh38
rs185234137 Chr.2: 202773369-202773369 on Build GRCh38
rs115739423 Chr.2: 202773056-202773056 on Build GRCh38
rs985765667 Chr.2: 202773441-202773441 on Build GRCh38
rs897260938 Chr.2: 202773197-202773197 on Build GRCh38
rs957761190 Chr.2: 202773434-202773434 on Build GRCh38
rs994136942 Chr.2: 202773254-202773255 on Build GRCh38
rs138893395 Chr.2: 202773046-202773046 on Build GRCh38
rs767389986 Chr.2: 202773397-202773397 on Build GRCh38
rs75494893 Chr.2: 202773262-202773262 on Build GRCh38

View Full Product Details