Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202773754-202774254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICA1L
Entrez Gene ID
130026
Gene Name
islet cell autoantigen 1 like
Gene Aliases
ALS2CR14, ALS2CR15
UniGene
Hs.516629
Gene Chromosome Location
Chr.2: 202773150-202871985 on Build GRCh38
Transcripts
NM_001288622.1 NM_001288623.1 NM_138468.5

SNP Detail

SNP ID Chromosome Location
rs137986703 Chr.2: 202773977-202773977 on Build GRCh38
rs896030434 Chr.2: 202773967-202773967 on Build GRCh38
rs965076415 Chr.2: 202774129-202774129 on Build GRCh38
rs530493159 Chr.2: 202774048-202774048 on Build GRCh38
rs903346326 Chr.2: 202774030-202774030 on Build GRCh38
rs564559979 Chr.2: 202774095-202774095 on Build GRCh38
rs186595866 Chr.2: 202774015-202774015 on Build GRCh38
rs543604665 Chr.2: 202774214-202774214 on Build GRCh38
rs911100303 Chr.2: 202773835-202773835 on Build GRCh38
rs539185450 Chr.2: 202773802-202773802 on Build GRCh38
rs549077983 Chr.2: 202774180-202774180 on Build GRCh38
rs976705729 Chr.2: 202773880-202773880 on Build GRCh38
rs181217247 Chr.2: 202773879-202773879 on Build GRCh38
rs769627525 Chr.2: 202774116-202774116 on Build GRCh38
rs547051664 Chr.2: 202773783-202773783 on Build GRCh38
rs957923180 Chr.2: 202773793-202773793 on Build GRCh38
rs918416507 Chr.2: 202773888-202773888 on Build GRCh38
rs529529592 Chr.2: 202774222-202774222 on Build GRCh38
rs535856677 Chr.2: 202773905-202773905 on Build GRCh38
rs538366580 Chr.2: 202774002-202774002 on Build GRCh38
rs781737532 Chr.2: 202774157-202774157 on Build GRCh38
rs562424155 Chr.2: 202774219-202774219 on Build GRCh38
rs937312026 Chr.2: 202773961-202773961 on Build GRCh38
rs1025590849 Chr.2: 202774148-202774148 on Build GRCh38
rs553911248 Chr.2: 202773962-202773962 on Build GRCh38
rs373905865 Chr.2: 202773794-202773794 on Build GRCh38
rs951173513 Chr.2: 202774154-202774154 on Build GRCh38
rs1000343598 Chr.2: 202774031-202774031 on Build GRCh38
rs909988550 Chr.2: 202773946-202773946 on Build GRCh38
rs150337227 Chr.2: 202773780-202773780 on Build GRCh38
rs551043197 Chr.2: 202773828-202773828 on Build GRCh38
rs929824589 Chr.2: 202773901-202773901 on Build GRCh38
rs1032120562 Chr.2: 202773787-202773791 on Build GRCh38
rs569561748 Chr.2: 202773842-202773842 on Build GRCh38
rs760847625 Chr.2: 202774126-202774126 on Build GRCh38
rs56411119 Chr.2: 202774065-202774065 on Build GRCh38
rs572356062 Chr.2: 202774133-202774136 on Build GRCh38
rs10870 Chr.2: 202774142-202774142 on Build GRCh38
rs367592195 Chr.2: 202774233-202774233 on Build GRCh38
rs569918137 Chr.2: 202773937-202773937 on Build GRCh38
rs573807601 Chr.2: 202773885-202773885 on Build GRCh38
rs751926610 Chr.2: 202773872-202773872 on Build GRCh38
rs1048942154 Chr.2: 202773903-202773903 on Build GRCh38
rs893569236 Chr.2: 202774066-202774066 on Build GRCh38
rs536839489 Chr.2: 202773866-202773866 on Build GRCh38
rs576717051 Chr.2: 202774123-202774123 on Build GRCh38
rs1017937748 Chr.2: 202774121-202774121 on Build GRCh38
rs965293500 Chr.2: 202773857-202773857 on Build GRCh38
rs776666885 Chr.2: 202774137-202774140 on Build GRCh38
rs748189466 Chr.2: 202774220-202774220 on Build GRCh38

View Full Product Details