Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202774499-202775017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICA1L
Entrez Gene ID
130026
Gene Name
islet cell autoantigen 1 like
Gene Aliases
ALS2CR14, ALS2CR15
UniGene
Hs.516629
Gene Chromosome Location
Chr.2: 202773150-202871985 on Build GRCh38
Transcripts
NM_001288622.1 NM_001288623.1 NM_138468.5

SNP Detail

SNP ID Chromosome Location
rs1017292589 Chr.2: 202774636-202774636 on Build GRCh38
rs72932709 Chr.2: 202774778-202774778 on Build GRCh38
rs892463877 Chr.2: 202774971-202774971 on Build GRCh38
rs948187070 Chr.2: 202774538-202774538 on Build GRCh38
rs951724039 Chr.2: 202774839-202774839 on Build GRCh38
rs567438192 Chr.2: 202774610-202774610 on Build GRCh38
rs948260294 Chr.2: 202774898-202774898 on Build GRCh38
rs377169243 Chr.2: 202774758-202774758 on Build GRCh38
rs72932707 Chr.2: 202774672-202774672 on Build GRCh38
rs1003568496 Chr.2: 202774605-202774605 on Build GRCh38
rs1045300253 Chr.2: 202774552-202774552 on Build GRCh38
rs553229062 Chr.2: 202774745-202774745 on Build GRCh38
rs946704866 Chr.2: 202774978-202774978 on Build GRCh38
rs892437077 Chr.2: 202774658-202774658 on Build GRCh38
rs939610090 Chr.2: 202774938-202774938 on Build GRCh38
rs537255430 Chr.2: 202774526-202774526 on Build GRCh38
rs771998735 Chr.2: 202774944-202774944 on Build GRCh38
rs1046879043 Chr.2: 202774875-202774875 on Build GRCh38
rs1005652340 Chr.2: 202774912-202774912 on Build GRCh38
rs1043867423 Chr.2: 202774979-202774979 on Build GRCh38
rs1018912679 Chr.2: 202774776-202774776 on Build GRCh38
rs933726689 Chr.2: 202774852-202774852 on Build GRCh38
rs990399501 Chr.2: 202774783-202774783 on Build GRCh38
rs1046769644 Chr.2: 202774550-202774550 on Build GRCh38
rs749494005 Chr.2: 202774659-202774659 on Build GRCh38
rs576576098 Chr.2: 202774861-202774861 on Build GRCh38
rs548869607 Chr.2: 202774572-202774572 on Build GRCh38
rs1024315059 Chr.2: 202774700-202774701 on Build GRCh38
rs1052851348 Chr.2: 202774642-202774642 on Build GRCh38
rs143905411 Chr.2: 202774848-202774848 on Build GRCh38
rs980885751 Chr.2: 202774906-202774906 on Build GRCh38
rs1026414438 Chr.2: 202774823-202774823 on Build GRCh38
rs868804291 Chr.2: 202774600-202774600 on Build GRCh38
rs971786371 Chr.2: 202774784-202774784 on Build GRCh38
rs943523505 Chr.2: 202774789-202774789 on Build GRCh38
rs1022348395 Chr.2: 202774675-202774675 on Build GRCh38
rs915629650 Chr.2: 202774894-202774894 on Build GRCh38
rs901350380 Chr.2: 202774772-202774772 on Build GRCh38
rs886816165 Chr.2: 202774580-202774580 on Build GRCh38
rs997819427 Chr.2: 202774542-202774542 on Build GRCh38
rs928246353 Chr.2: 202774908-202774908 on Build GRCh38

View Full Product Details