Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202776490-202777012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICA1L
Entrez Gene ID
130026
Gene Name
islet cell autoantigen 1 like
Gene Aliases
ALS2CR14, ALS2CR15
UniGene
Hs.516629
Gene Chromosome Location
Chr.2: 202773150-202871985 on Build GRCh38
Transcripts
NM_001288622.1 NM_001288623.1 NM_138468.5

SNP Detail

SNP ID Chromosome Location
rs747421981 Chr.2: 202776737-202776737 on Build GRCh38
rs546684739 Chr.2: 202776954-202776954 on Build GRCh38
rs542559897 Chr.2: 202776639-202776639 on Build GRCh38
rs772661581 Chr.2: 202776660-202776660 on Build GRCh38
rs997798573 Chr.2: 202776738-202776738 on Build GRCh38
rs909288907 Chr.2: 202776565-202776565 on Build GRCh38
rs979903570 Chr.2: 202776598-202776598 on Build GRCh38
rs532346833 Chr.2: 202776829-202776829 on Build GRCh38
rs368977062 Chr.2: 202776541-202776541 on Build GRCh38
rs569271730 Chr.2: 202776643-202776643 on Build GRCh38
rs950777918 Chr.2: 202776807-202776807 on Build GRCh38
rs927131176 Chr.2: 202776869-202776869 on Build GRCh38
rs973931746 Chr.2: 202776971-202776971 on Build GRCh38
rs1057398375 Chr.2: 202776651-202776651 on Build GRCh38
rs891548923 Chr.2: 202776814-202776814 on Build GRCh38
rs530814114 Chr.2: 202776870-202776870 on Build GRCh38
rs150657239 Chr.2: 202776815-202776815 on Build GRCh38
rs927247387 Chr.2: 202776606-202776606 on Build GRCh38
rs760043334 Chr.2: 202776516-202776516 on Build GRCh38
rs771109763 Chr.2: 202776787-202776787 on Build GRCh38
rs142926699 Chr.2: 202776673-202776673 on Build GRCh38
rs193071238 Chr.2: 202776945-202776945 on Build GRCh38
rs938619862 Chr.2: 202776638-202776638 on Build GRCh38
rs940784945 Chr.2: 202776962-202776962 on Build GRCh38
rs904253865 Chr.2: 202776683-202776683 on Build GRCh38
rs987533960 Chr.2: 202776548-202776548 on Build GRCh38
rs974643721 Chr.2: 202776866-202776866 on Build GRCh38
rs561033099 Chr.2: 202776827-202776827 on Build GRCh38
rs945672776 Chr.2: 202776662-202776662 on Build GRCh38
rs113452458 Chr.2: 202776676-202776676 on Build GRCh38
rs1028937754 Chr.2: 202776875-202776875 on Build GRCh38
rs941893390 Chr.2: 202776566-202776566 on Build GRCh38
rs913126321 Chr.2: 202776919-202776919 on Build GRCh38
rs188124932 Chr.2: 202776914-202776914 on Build GRCh38
rs374051844 Chr.2: 202776724-202776724 on Build GRCh38
rs1017776438 Chr.2: 202776707-202776707 on Build GRCh38
rs1001071163 Chr.2: 202776703-202776703 on Build GRCh38
rs1035372103 Chr.2: 202776792-202776792 on Build GRCh38
rs907967269 Chr.2: 202776575-202776575 on Build GRCh38
rs183339208 Chr.2: 202776839-202776839 on Build GRCh38
rs191677391 Chr.2: 202776608-202776608 on Build GRCh38
rs1030973059 Chr.2: 202776782-202776782 on Build GRCh38
rs116272690 Chr.2: 202776844-202776844 on Build GRCh38
rs933167622 Chr.2: 202776544-202776544 on Build GRCh38
rs567670948 Chr.2: 202776917-202776917 on Build GRCh38
rs907912550 Chr.2: 202776958-202776958 on Build GRCh38

View Full Product Details