The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014314.3 | NP_055129.2 | 2 - 3 | 398 | 76 |
GenBank mRNA | AF038963.1 | - | 2 - 3 | 397 | 76 |
AK301678.1 | - | 2 - 3 | 269 | 76 | |
AK315040.1 | - | 2 - 3 | 269 | 76 | |
BC107731.1 | - | 2 - 3 | 420 | 76 | |
BC132786.1 | - | 2 - 3 | 346 | 76 | |
BC136610.1 | - | 2 - 3 | 346 | 76 | |
DN998145.1 | - | 2 - 3 | 399 | 76 |