Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 203366785-203367294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABI2
Entrez Gene ID
10152
Gene Name
abl-interactor 2
Gene Aliases
ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
UniGene
Hs.471156
Gene Chromosome Location
Chr.2: 203327874-203432173 on Build GRCh38
Transcripts
NM_001282925.1 NM_001282926.1 NM_001282927.1 NM_001282932.1 NM_005759.5 XM_005246217.2 XM_005246218.2 XM_005246221.2 XM_005246225.2 XM_005246226.2 XM_006712174.2 XM_006712177.2 XM_006712179.2 XM_006712180.2 XM_006712181.2 XM_006712182.2 XM_006712185.1 XM_006712186.2 XM_011510472.2 XM_011510473.2 XM_017003118.1 XM_017003119.1 XM_017003120.1 XM_017003121.1 XM_017003122.1 XM_017003123.1 XM_017003124.1 XM_017003125.1 XM_017003126.1 XM_017003127.1 XM_017003128.1 XM_017003129.1 XM_017003130.1 XM_017003131.1 XM_017003132.1 XM_017003133.1 XM_017003134.1 XM_017003135.1 XM_017003136.1 XM_017003137.1 XM_017003138.1 XM_017003139.1 XM_017003140.1 XM_017003141.1 XM_017003142.1 XM_017003143.1 XM_017003144.1 XM_017003145.1 XM_017003146.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7316416 c.285+1G>C p.? Chr.2: 203367045-203367045 on Build GRCh38
5806083 c.259G>T p.E87* Chr.2: 203367018-203367018 on Build GRCh38
6266158 c.214G>T p.V72F Chr.2: 203366973-203366973 on Build GRCh38
7105625 c.237G>A p.Q79Q Chr.2: 203366996-203366996 on Build GRCh38
3576504 c.171C>T p.S57S Chr.2: 203366930-203366930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773240579 Chr.2: 203367093-203367093 on Build GRCh38
rs773980418 Chr.2: 203366896-203366897 on Build GRCh38
rs989241706 Chr.2: 203367206-203367206 on Build GRCh38
rs756857928 Chr.2: 203366852-203366852 on Build GRCh38
rs1003463669 Chr.2: 203367174-203367174 on Build GRCh38
rs201137565 Chr.2: 203367088-203367088 on Build GRCh38
rs376386538 Chr.2: 203366834-203366834 on Build GRCh38
rs998123243 Chr.2: 203367235-203367235 on Build GRCh38
rs887115193 Chr.2: 203367098-203367098 on Build GRCh38
rs1056362475 Chr.2: 203367104-203367104 on Build GRCh38
rs778355722 Chr.2: 203366864-203366864 on Build GRCh38
rs749300109 Chr.2: 203367071-203367071 on Build GRCh38
rs558177158 Chr.2: 203367141-203367141 on Build GRCh38
rs914996956 Chr.2: 203367214-203367214 on Build GRCh38
rs887831204 Chr.2: 203367153-203367153 on Build GRCh38
rs749102478 Chr.2: 203366881-203366881 on Build GRCh38
rs754283915 Chr.2: 203366972-203366972 on Build GRCh38
rs768680496 Chr.2: 203367074-203367074 on Build GRCh38
rs780661656 Chr.2: 203367051-203367051 on Build GRCh38
rs780103534 Chr.2: 203366871-203366871 on Build GRCh38
rs769877930 Chr.2: 203366858-203366858 on Build GRCh38
rs765659242 Chr.2: 203367015-203367015 on Build GRCh38
rs755268731 Chr.2: 203366847-203366847 on Build GRCh38
rs1005561183 Chr.2: 203367101-203367101 on Build GRCh38
rs1015056471 Chr.2: 203366891-203366891 on Build GRCh38
rs752975781 Chr.2: 203367027-203367027 on Build GRCh38
rs970605240 Chr.2: 203367158-203367158 on Build GRCh38
rs749984907 Chr.2: 203367052-203367052 on Build GRCh38
rs780156520 Chr.2: 203367066-203367066 on Build GRCh38
rs749839606 Chr.2: 203366871-203366872 on Build GRCh38
rs778815155 Chr.2: 203367082-203367082 on Build GRCh38
rs756558806 Chr.2: 203366863-203366864 on Build GRCh38
rs1012519430 Chr.2: 203367121-203367121 on Build GRCh38
rs989642946 Chr.2: 203366918-203366918 on Build GRCh38
rs770512647 Chr.2: 203366892-203366893 on Build GRCh38
rs745349093 Chr.2: 203366857-203366857 on Build GRCh38
rs554617778 Chr.2: 203367096-203367096 on Build GRCh38
rs1004939017 Chr.2: 203367193-203367193 on Build GRCh38
rs772158653 Chr.2: 203366921-203366921 on Build GRCh38
rs754231144 Chr.2: 203366836-203366836 on Build GRCh38
rs762133397 Chr.2: 203366915-203366915 on Build GRCh38
rs751285843 Chr.2: 203366851-203366851 on Build GRCh38
rs956393230 Chr.2: 203367192-203367195 on Build GRCh38
rs966164479 Chr.2: 203367219-203367219 on Build GRCh38
rs1030637284 Chr.2: 203367191-203367191 on Build GRCh38
rs966239769 Chr.2: 203366931-203366931 on Build GRCh38
rs953725860 Chr.2: 203366959-203366959 on Build GRCh38
rs755623364 Chr.2: 203367054-203367054 on Build GRCh38
rs747845264 Chr.2: 203367083-203367083 on Build GRCh38
rs922201744 Chr.2: 203366938-203366938 on Build GRCh38
rs533498296 Chr.2: 203367128-203367128 on Build GRCh38
rs774280959 Chr.2: 203366898-203366898 on Build GRCh38
rs1012249510 Chr.2: 203367129-203367129 on Build GRCh38
rs773360186 Chr.2: 203366942-203366942 on Build GRCh38
rs985564559 Chr.2: 203366966-203366966 on Build GRCh38
rs563394530 Chr.2: 203366900-203366900 on Build GRCh38
rs766875319 Chr.2: 203366960-203366960 on Build GRCh38
rs942900841 Chr.2: 203367041-203367041 on Build GRCh38
rs981803208 Chr.2: 203366814-203366814 on Build GRCh38
rs1029239922 Chr.2: 203367246-203367246 on Build GRCh38
rs976381160 Chr.2: 203366936-203366936 on Build GRCh38
rs760630657 Chr.2: 203366957-203366957 on Build GRCh38
rs777414061 Chr.2: 203366849-203366849 on Build GRCh38
rs1015023861 Chr.2: 203367216-203367216 on Build GRCh38
rs761796222 Chr.2: 203367010-203367010 on Build GRCh38
rs975956542 Chr.2: 203367240-203367240 on Build GRCh38
rs566241233 Chr.2: 203367107-203367107 on Build GRCh38
rs1023540096 Chr.2: 203367150-203367150 on Build GRCh38
rs756983744 Chr.2: 203367031-203367031 on Build GRCh38
rs35533326 Chr.2: 203367212-203367212 on Build GRCh38
rs768516875 Chr.2: 203366894-203366894 on Build GRCh38
rs923782180 Chr.2: 203367081-203367081 on Build GRCh38
rs780795206 Chr.2: 203366854-203366854 on Build GRCh38

View Full Product Details