Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 203380056-203380556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABI2
Entrez Gene ID
10152
Gene Name
abl-interactor 2
Gene Aliases
ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
UniGene
Hs.471156
Gene Chromosome Location
Chr.2: 203327874-203432173 on Build GRCh38
Transcripts
NM_001282925.1 NM_001282926.1 NM_001282927.1 NM_001282932.1 NM_005759.5 XM_005246217.2 XM_005246218.2 XM_005246221.2 XM_005246225.2 XM_005246226.2 XM_006712174.2 XM_006712177.2 XM_006712179.2 XM_006712180.2 XM_006712181.2 XM_006712182.2 XM_006712185.1 XM_006712186.2 XM_011510472.2 XM_011510473.2 XM_017003118.1 XM_017003119.1 XM_017003120.1 XM_017003121.1 XM_017003122.1 XM_017003123.1 XM_017003124.1 XM_017003125.1 XM_017003126.1 XM_017003127.1 XM_017003128.1 XM_017003129.1 XM_017003130.1 XM_017003131.1 XM_017003132.1 XM_017003133.1 XM_017003134.1 XM_017003135.1 XM_017003136.1 XM_017003137.1 XM_017003138.1 XM_017003139.1 XM_017003140.1 XM_017003141.1 XM_017003142.1 XM_017003143.1 XM_017003144.1 XM_017003145.1 XM_017003146.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5725605 c.237T>G p.I79M Chr.2: 203380294-203380294 on Build GRCh38
5708011 c.287C>A p.T96K Chr.2: 203380209-203380209 on Build GRCh38
5725604 c.372T>G p.I124M Chr.2: 203380294-203380294 on Build GRCh38
1015121 c.322G>T p.E108* Chr.2: 203380244-203380244 on Build GRCh38
5435035 c.388C>A p.L130I Chr.2: 203380310-203380310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758351931 Chr.2: 203380411-203380411 on Build GRCh38
rs774776788 Chr.2: 203380331-203380331 on Build GRCh38
rs528637435 Chr.2: 203380128-203380128 on Build GRCh38
rs549599768 Chr.2: 203380197-203380197 on Build GRCh38
rs369306670 Chr.2: 203380259-203380259 on Build GRCh38
rs774449659 Chr.2: 203380524-203380524 on Build GRCh38
rs911082572 Chr.2: 203380210-203380210 on Build GRCh38
rs1030554672 Chr.2: 203380496-203380496 on Build GRCh38
rs754983755 Chr.2: 203380206-203380206 on Build GRCh38
rs1033489407 Chr.2: 203380155-203380155 on Build GRCh38
rs965287005 Chr.2: 203380283-203380283 on Build GRCh38
rs905220620 Chr.2: 203380352-203380352 on Build GRCh38
rs745899975 Chr.2: 203380290-203380290 on Build GRCh38
rs753426864 Chr.2: 203380358-203380358 on Build GRCh38
rs1048077664 Chr.2: 203380400-203380400 on Build GRCh38
rs777468584 Chr.2: 203380420-203380420 on Build GRCh38
rs1043485811 Chr.2: 203380131-203380131 on Build GRCh38
rs550137591 Chr.2: 203380469-203380469 on Build GRCh38
rs765232686 Chr.2: 203380395-203380395 on Build GRCh38
rs546922774 Chr.2: 203380141-203380141 on Build GRCh38
rs1006299740 Chr.2: 203380376-203380376 on Build GRCh38
rs910952407 Chr.2: 203380087-203380087 on Build GRCh38
rs942436838 Chr.2: 203380096-203380096 on Build GRCh38
rs766075623 Chr.2: 203380350-203380350 on Build GRCh38
rs754467735 Chr.2: 203380368-203380368 on Build GRCh38
rs903688102 Chr.2: 203380140-203380140 on Build GRCh38
rs761761705 Chr.2: 203380426-203380427 on Build GRCh38
rs896406791 Chr.2: 203380295-203380295 on Build GRCh38
rs967815075 Chr.2: 203380124-203380124 on Build GRCh38
rs561574948 Chr.2: 203380107-203380107 on Build GRCh38
rs776742630 Chr.2: 203380282-203380282 on Build GRCh38
rs755817692 Chr.2: 203380170-203380170 on Build GRCh38
rs574221820 Chr.2: 203380425-203380425 on Build GRCh38
rs764426893 Chr.2: 203380330-203380330 on Build GRCh38
rs370999877 Chr.2: 203380421-203380421 on Build GRCh38
rs775797737 Chr.2: 203380322-203380322 on Build GRCh38
rs747978075 Chr.2: 203380217-203380217 on Build GRCh38
rs563435995 Chr.2: 203380080-203380080 on Build GRCh38
rs747297124 Chr.2: 203380272-203380272 on Build GRCh38
rs141392713 Chr.2: 203380343-203380343 on Build GRCh38
rs1802776 Chr.2: 203380310-203380310 on Build GRCh38
rs372540709 Chr.2: 203380184-203380184 on Build GRCh38
rs1055868970 Chr.2: 203380518-203380518 on Build GRCh38
rs776511177 Chr.2: 203380409-203380410 on Build GRCh38
rs760330350 Chr.2: 203380185-203380186 on Build GRCh38
rs1014480402 Chr.2: 203380104-203380104 on Build GRCh38
rs962163951 Chr.2: 203380429-203380429 on Build GRCh38
rs778671701 Chr.2: 203380212-203380212 on Build GRCh38
rs763883143 Chr.2: 203380392-203380407 on Build GRCh38
rs771111985 Chr.2: 203380281-203380281 on Build GRCh38
rs752632496 Chr.2: 203380406-203380406 on Build GRCh38
rs777608395 Chr.2: 203380266-203380266 on Build GRCh38
rs770364969 Chr.2: 203380320-203380320 on Build GRCh38
rs909606291 Chr.2: 203380434-203380434 on Build GRCh38
rs371242683 Chr.2: 203380185-203380185 on Build GRCh38
rs763393954 Chr.2: 203380328-203380328 on Build GRCh38
rs370812418 Chr.2: 203380157-203380157 on Build GRCh38
rs1000205485 Chr.2: 203380137-203380137 on Build GRCh38
rs549666473 Chr.2: 203380095-203380095 on Build GRCh38

View Full Product Details