Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 203734705-203735217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD28
Entrez Gene ID
940
Gene Name
CD28 molecule
Gene Aliases
Tp44
UniGene
Hs.443123
Gene Chromosome Location
Chr.2: 203706475-203739756 on Build GRCh38
Transcripts
NM_001243077.1 NM_001243078.1 NM_006139.3 XM_011512194.2 XM_011512195.2 XM_011512197.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5460804 c.633A>C p.P211P Chr.2: 203734882-203734882 on Build GRCh38
6847386 c.587C>A p.P196H Chr.2: 203734836-203734836 on Build GRCh38
243564 c.593G>A p.R198H Chr.2: 203734842-203734842 on Build GRCh38
3838358 c.655C>T p.R219C Chr.2: 203734904-203734904 on Build GRCh38
2157254 c.589C>T p.R197C Chr.2: 203734838-203734838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200921638 Chr.2: 203735096-203735096 on Build GRCh38
rs139881881 Chr.2: 203734809-203734809 on Build GRCh38
rs745917782 Chr.2: 203734773-203734773 on Build GRCh38
rs77341727 Chr.2: 203735126-203735126 on Build GRCh38
rs200016310 Chr.2: 203734895-203734895 on Build GRCh38
rs896281170 Chr.2: 203734774-203734774 on Build GRCh38
rs201180139 Chr.2: 203734748-203734748 on Build GRCh38
rs367908475 Chr.2: 203734856-203734856 on Build GRCh38
rs145761335 Chr.2: 203734841-203734841 on Build GRCh38
rs371010635 Chr.2: 203734747-203734747 on Build GRCh38
rs201285390 Chr.2: 203735146-203735146 on Build GRCh38
rs763060776 Chr.2: 203734980-203734980 on Build GRCh38
rs201159081 Chr.2: 203734920-203734920 on Build GRCh38
rs368916983 Chr.2: 203734919-203734919 on Build GRCh38
rs202064511 Chr.2: 203734928-203734928 on Build GRCh38
rs201499339 Chr.2: 203735056-203735056 on Build GRCh38
rs896816560 Chr.2: 203734737-203734737 on Build GRCh38
rs201276180 Chr.2: 203734938-203734938 on Build GRCh38
rs531576854 Chr.2: 203734807-203734807 on Build GRCh38
rs202151959 Chr.2: 203735158-203735158 on Build GRCh38
rs201547332 Chr.2: 203734848-203734848 on Build GRCh38
rs763432798 Chr.2: 203734784-203734784 on Build GRCh38
rs771164270 Chr.2: 203734767-203734767 on Build GRCh38
rs199549636 Chr.2: 203734857-203734857 on Build GRCh38
rs200922648 Chr.2: 203735058-203735058 on Build GRCh38
rs562969933 Chr.2: 203734832-203734832 on Build GRCh38
rs78722979 Chr.2: 203735128-203735128 on Build GRCh38
rs201909740 Chr.2: 203734889-203734889 on Build GRCh38
rs749688881 Chr.2: 203734862-203734862 on Build GRCh38
rs138598762 Chr.2: 203735174-203735174 on Build GRCh38
rs751945323 Chr.2: 203734812-203734812 on Build GRCh38
rs767908312 Chr.2: 203735047-203735047 on Build GRCh38
rs1013433065 Chr.2: 203734785-203734785 on Build GRCh38
rs200642723 Chr.2: 203734850-203734850 on Build GRCh38
rs757690722 Chr.2: 203735029-203735029 on Build GRCh38
rs188032362 Chr.2: 203734813-203734813 on Build GRCh38
rs200119144 Chr.2: 203735093-203735093 on Build GRCh38
rs570801289 Chr.2: 203734926-203734926 on Build GRCh38
rs200538326 Chr.2: 203734933-203734933 on Build GRCh38
rs371850110 Chr.2: 203734874-203734874 on Build GRCh38
rs924918302 Chr.2: 203734989-203734989 on Build GRCh38
rs368834988 Chr.2: 203735184-203735184 on Build GRCh38
rs80306002 Chr.2: 203735088-203735088 on Build GRCh38
rs749826156 Chr.2: 203734925-203734925 on Build GRCh38
rs200167780 Chr.2: 203735173-203735173 on Build GRCh38
rs911179223 Chr.2: 203735192-203735192 on Build GRCh38
rs140455620 Chr.2: 203734913-203734913 on Build GRCh38
rs1023502670 Chr.2: 203734838-203734838 on Build GRCh38
rs572054130 Chr.2: 203735031-203735031 on Build GRCh38
rs370228563 Chr.2: 203734903-203734903 on Build GRCh38
rs576354946 Chr.2: 203735149-203735149 on Build GRCh38
rs1030322547 Chr.2: 203735187-203735187 on Build GRCh38
rs199784948 Chr.2: 203734978-203734978 on Build GRCh38
rs747144866 Chr.2: 203734761-203734761 on Build GRCh38
rs960188485 Chr.2: 203735014-203735014 on Build GRCh38
rs201899664 Chr.2: 203735074-203735074 on Build GRCh38
rs770610915 Chr.2: 203734883-203734883 on Build GRCh38
rs200161233 Chr.2: 203734990-203734990 on Build GRCh38
rs34935465 Chr.2: 203735013-203735013 on Build GRCh38
rs199912903 Chr.2: 203734916-203734916 on Build GRCh38
rs200552329 Chr.2: 203735085-203735085 on Build GRCh38
rs200606770 Chr.2: 203734871-203734871 on Build GRCh38
rs199979718 Chr.2: 203735110-203735110 on Build GRCh38
rs770459409 Chr.2: 203734776-203734776 on Build GRCh38
rs764067537 Chr.2: 203734912-203734912 on Build GRCh38
rs373620796 Chr.2: 203734914-203734914 on Build GRCh38
rs1052320187 Chr.2: 203734997-203734997 on Build GRCh38
rs967467536 Chr.2: 203735069-203735069 on Build GRCh38
rs762144222 Chr.2: 203734881-203734881 on Build GRCh38
rs201673371 Chr.2: 203734915-203734915 on Build GRCh38
rs1020345772 Chr.2: 203735064-203735064 on Build GRCh38
rs201902842 Chr.2: 203735099-203735099 on Build GRCh38
rs574216169 Chr.2: 203734959-203734959 on Build GRCh38
rs762747357 Chr.2: 203734908-203734908 on Build GRCh38
rs1016664157 Chr.2: 203734854-203734854 on Build GRCh38
rs200289442 Chr.2: 203734953-203734953 on Build GRCh38
rs752011627 Chr.2: 203735029-203735029 on Build GRCh38
rs748579255 Chr.2: 203734951-203734951 on Build GRCh38
rs57861808 Chr.2: 203735044-203735044 on Build GRCh38
rs143458715 Chr.2: 203734751-203734751 on Build GRCh38
rs199777674 Chr.2: 203734886-203734886 on Build GRCh38
rs116310870 Chr.2: 203734846-203734846 on Build GRCh38
rs910759207 Chr.2: 203734946-203734946 on Build GRCh38
rs201423059 Chr.2: 203734960-203734960 on Build GRCh38
rs200751829 Chr.2: 203734847-203734847 on Build GRCh38
rs200936737 Chr.2: 203734904-203734904 on Build GRCh38
rs936603709 Chr.2: 203734873-203734873 on Build GRCh38
rs776800736 Chr.2: 203734772-203734772 on Build GRCh38
rs201991178 Chr.2: 203734939-203734939 on Build GRCh38
rs985772089 Chr.2: 203735183-203735183 on Build GRCh38
rs200221759 Chr.2: 203734829-203734829 on Build GRCh38
rs769098383 Chr.2: 203734863-203734863 on Build GRCh38
rs535570029 Chr.2: 203734971-203734971 on Build GRCh38
rs199647272 Chr.2: 203734842-203734842 on Build GRCh38
rs753396357 Chr.2: 203734814-203734814 on Build GRCh38
rs199553976 Chr.2: 203735082-203735082 on Build GRCh38
rs142698532 Chr.2: 203734888-203734888 on Build GRCh38
rs774531956 Chr.2: 203734870-203734870 on Build GRCh38
rs573551913 Chr.2: 203735102-203735102 on Build GRCh38
rs897791387 Chr.2: 203735157-203735157 on Build GRCh38
rs192635437 Chr.2: 203735046-203735046 on Build GRCh38
rs201598596 Chr.2: 203734905-203734905 on Build GRCh38
rs201773411 Chr.2: 203734878-203734878 on Build GRCh38
rs888683386 Chr.2: 203734948-203734948 on Build GRCh38
rs572738990 Chr.2: 203734866-203734866 on Build GRCh38
rs754453810 Chr.2: 203734824-203734824 on Build GRCh38
rs775856796 Chr.2: 203734780-203734780 on Build GRCh38
rs199643331 Chr.2: 203735057-203735057 on Build GRCh38
rs778118170 Chr.2: 203734754-203734754 on Build GRCh38
rs765515314 Chr.2: 203734839-203734839 on Build GRCh38
rs1037806138 Chr.2: 203735121-203735121 on Build GRCh38
rs374028740 Chr.2: 203734781-203734781 on Build GRCh38
rs778799984 Chr.2: 203734746-203734746 on Build GRCh38

View Full Product Details