Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 206543169-206543618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM23
Entrez Gene ID
8745
Gene Name
ADAM metallopeptidase domain 23
Gene Aliases
MDC-3, MDC3
UniGene
Hs.370287
Gene Chromosome Location
Chr.2: 206443544-206621130 on Build GRCh38
Transcripts
NM_003812.3 XM_005246932.3 XM_011512086.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6090428 c.658G>C p.G220R Chr.2: 206543254-206543254 on Build GRCh38
7157816 c.720G>A p.E240E Chr.2: 206543316-206543316 on Build GRCh38
5669644 c.657-3T>C p.? Chr.2: 206543250-206543250 on Build GRCh38
6090430 c.658G>C p.G220R Chr.2: 206543254-206543254 on Build GRCh38
7157817 c.720G>A p.E240E Chr.2: 206543316-206543316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370855019 Chr.2: 206543221-206543221 on Build GRCh38
rs1044594275 Chr.2: 206543234-206543234 on Build GRCh38
rs1034295179 Chr.2: 206543309-206543309 on Build GRCh38
rs762262591 Chr.2: 206543497-206543497 on Build GRCh38
rs540935024 Chr.2: 206543400-206543400 on Build GRCh38
rs1016345412 Chr.2: 206543410-206543410 on Build GRCh38
rs747379471 Chr.2: 206543237-206543237 on Build GRCh38
rs754075810 Chr.2: 206543304-206543304 on Build GRCh38
rs755129983 Chr.2: 206543323-206543323 on Build GRCh38
rs954880741 Chr.2: 206543456-206543456 on Build GRCh38
rs1030491333 Chr.2: 206543447-206543447 on Build GRCh38
rs374039354 Chr.2: 206543485-206543485 on Build GRCh38
rs775278833 Chr.2: 206543257-206543257 on Build GRCh38
rs746412949 Chr.2: 206543537-206543537 on Build GRCh38
rs192110552 Chr.2: 206543523-206543523 on Build GRCh38
rs994575171 Chr.2: 206543311-206543311 on Build GRCh38
rs748610387 Chr.2: 206543220-206543220 on Build GRCh38
rs949815529 Chr.2: 206543504-206543504 on Build GRCh38
rs544096354 Chr.2: 206543412-206543412 on Build GRCh38
rs187664975 Chr.2: 206543494-206543494 on Build GRCh38
rs938495027 Chr.2: 206543550-206543550 on Build GRCh38
rs199976489 Chr.2: 206543227-206543227 on Build GRCh38
rs139914873 Chr.2: 206543524-206543524 on Build GRCh38
rs369026982 Chr.2: 206543329-206543329 on Build GRCh38
rs905661681 Chr.2: 206543518-206543518 on Build GRCh38
rs376726273 Chr.2: 206543325-206543325 on Build GRCh38
rs955417537 Chr.2: 206543555-206543555 on Build GRCh38
rs376065204 Chr.2: 206543339-206543339 on Build GRCh38
rs768286271 Chr.2: 206543270-206543270 on Build GRCh38
rs569819538 Chr.2: 206543574-206543574 on Build GRCh38
rs1030046726 Chr.2: 206543546-206543546 on Build GRCh38
rs143684302 Chr.2: 206543441-206543441 on Build GRCh38
rs146616400 Chr.2: 206543282-206543282 on Build GRCh38
rs756441845 Chr.2: 206543544-206543544 on Build GRCh38
rs147218303 Chr.2: 206543496-206543496 on Build GRCh38
rs182885374 Chr.2: 206543527-206543527 on Build GRCh38
rs112810207 Chr.2: 206543459-206543459 on Build GRCh38
rs905712752 Chr.2: 206543246-206543246 on Build GRCh38
rs986720500 Chr.2: 206543469-206543469 on Build GRCh38
rs955627061 Chr.2: 206543351-206543351 on Build GRCh38
rs762635513 Chr.2: 206543258-206543258 on Build GRCh38
rs1016870076 Chr.2: 206543289-206543289 on Build GRCh38
rs752934422 Chr.2: 206543272-206543272 on Build GRCh38
rs758879172 Chr.2: 206543368-206543368 on Build GRCh38
rs563788241 Chr.2: 206543420-206543420 on Build GRCh38
rs780464447 Chr.2: 206543549-206543549 on Build GRCh38
rs924186025 Chr.2: 206543490-206543490 on Build GRCh38
rs1044172964 Chr.2: 206543505-206543505 on Build GRCh38
rs531348178 Chr.2: 206543437-206543437 on Build GRCh38
rs141617022 Chr.2: 206543300-206543300 on Build GRCh38
rs775313540 Chr.2: 206543365-206543370 on Build GRCh38
rs1018319802 Chr.2: 206543561-206543561 on Build GRCh38
rs752776209 Chr.2: 206543347-206543347 on Build GRCh38
rs935636477 Chr.2: 206543491-206543491 on Build GRCh38
rs199943814 Chr.2: 206543278-206543278 on Build GRCh38
rs760455197 Chr.2: 206543285-206543285 on Build GRCh38
rs558319181 Chr.2: 206543277-206543277 on Build GRCh38
rs1056048391 Chr.2: 206543582-206543582 on Build GRCh38
rs368278452 Chr.2: 206543522-206543522 on Build GRCh38
rs778910355 Chr.2: 206543203-206543203 on Build GRCh38
rs1007345096 Chr.2: 206543558-206543558 on Build GRCh38
rs755041674 Chr.2: 206543202-206543202 on Build GRCh38
rs764252291 Chr.2: 206543483-206543483 on Build GRCh38
rs781735039 Chr.2: 206543509-206543509 on Build GRCh38

View Full Product Details