The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134335.1 | NP_001127807.1 | 1 - 2 | 259 | 74 |
NM_014413.3 | NP_055228.2 | 1 - 2 | 259 | 74 | |
GenBank mRNA | AB037790.1 | - | 1 - 2 | 184 | 74 |
AF100784.1 | - | 1 - 2 | 148 | 74 | |
AF147094.1 | - | 1 - 2 | 194 | 74 | |
AF183414.1 | - | 1 - 2 | 146 | 74 | |
AK075192.1 | - | 1 - 2 | 187 | 74 | |
AK290327.1 | - | 1 - 2 | 192 | 74 | |
AK295712.1 | - | 1 - 2 | 194 | 74 | |
AL136563.1 | - | 1 - 2 | 164 | 74 | |
AL834494.1 | - | 1 - 2 | 164 | 74 | |
BC006524.2 | - | 1 - 2 | 191 | 74 | |
DC341983.1 | - | 1 - 2 | 259 | 74 |