The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014456.4 | NP_055271.2 | 2 - 3 | 330 | 94 |
GenBank mRNA | AK223623.1 | - | 2 - 3 | 204 | 94 |
AK312540.1 | - | 2 - 3 | 226 | 94 | |
BC015036.2 | - | 2 - 3 | 194 | 94 | |
BC026104.2 | - | 2 - 3 | 197 | 94 | |
BC031049.1 | - | 2 - 3 | 197 | 94 | |
BI910668.1 | - | 2 - 3 | 292 | 94 | |
BX537500.1 | - | 1 - 2 | 3278 | 94 | |
DB241608.1 | - | 2 - 3 | 331 | 94 | |
U96628.1 | - | 2 - 3 | 128 | 94 |