Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218265974-218266463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAMP
Entrez Gene ID
14
Gene Name
angio associated migratory cell protein
Gene Aliases
-
UniGene
Hs.83347
Gene Chromosome Location
Chr.2: 218264129-218270209 on Build GRCh38
Transcripts
NM_001087.4 NM_001302545.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5565125 c.719G>A p.R240K Chr.2: 218266108-218266108 on Build GRCh38
79121 c.760A>T p.K254* Chr.2: 218266067-218266067 on Build GRCh38
1405444 c.664C>T p.R222* Chr.2: 218266458-218266458 on Build GRCh38
6337674 c.673C>A p.P225T Chr.2: 218266449-218266449 on Build GRCh38
4415485 c.748A>G p.I250V Chr.2: 218266079-218266079 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372583826 Chr.2: 218266082-218266082 on Build GRCh38
rs780596182 Chr.2: 218266173-218266173 on Build GRCh38
rs932282857 Chr.2: 218266093-218266093 on Build GRCh38
rs148721412 Chr.2: 218266071-218266071 on Build GRCh38
rs377002351 Chr.2: 218266163-218266163 on Build GRCh38
rs1013182584 Chr.2: 218266270-218266270 on Build GRCh38
rs889295903 Chr.2: 218266045-218266045 on Build GRCh38
rs775271306 Chr.2: 218266197-218266197 on Build GRCh38
rs528748144 Chr.2: 218266038-218266038 on Build GRCh38
rs373583528 Chr.2: 218266398-218266398 on Build GRCh38
rs112909351 Chr.2: 218266400-218266400 on Build GRCh38
rs1000827608 Chr.2: 218266319-218266319 on Build GRCh38
rs374837832 Chr.2: 218266000-218266000 on Build GRCh38
rs756497659 Chr.2: 218266171-218266171 on Build GRCh38
rs935272254 Chr.2: 218266236-218266236 on Build GRCh38
rs879064483 Chr.2: 218266220-218266220 on Build GRCh38
rs569258969 Chr.2: 218266336-218266336 on Build GRCh38
rs763647016 Chr.2: 218266392-218266392 on Build GRCh38
rs922151100 Chr.2: 218266090-218266090 on Build GRCh38
rs1036038158 Chr.2: 218266235-218266235 on Build GRCh38
rs969211414 Chr.2: 218266206-218266206 on Build GRCh38
rs776528155 Chr.2: 218266192-218266192 on Build GRCh38
rs1050602811 Chr.2: 218266034-218266034 on Build GRCh38
rs558292653 Chr.2: 218266394-218266394 on Build GRCh38
rs369981705 Chr.2: 218266052-218266052 on Build GRCh38
rs757379596 Chr.2: 218266055-218266056 on Build GRCh38
rs777962923 Chr.2: 218266383-218266383 on Build GRCh38
rs896243057 Chr.2: 218266243-218266243 on Build GRCh38
rs979662846 Chr.2: 218266020-218266020 on Build GRCh38
rs372929094 Chr.2: 218266185-218266185 on Build GRCh38
rs762611900 Chr.2: 218266198-218266198 on Build GRCh38
rs778816857 Chr.2: 218266421-218266422 on Build GRCh38
rs763594720 Chr.2: 218266138-218266138 on Build GRCh38
rs756855084 Chr.2: 218266161-218266161 on Build GRCh38
rs2305836 Chr.2: 218266425-218266425 on Build GRCh38
rs534059484 Chr.2: 218266421-218266421 on Build GRCh38
rs1023285820 Chr.2: 218266300-218266300 on Build GRCh38
rs889244917 Chr.2: 218266293-218266293 on Build GRCh38
rs957682535 Chr.2: 218266046-218266046 on Build GRCh38
rs370369629 Chr.2: 218266167-218266167 on Build GRCh38
rs745832404 Chr.2: 218266193-218266193 on Build GRCh38
rs1050961984 Chr.2: 218266292-218266292 on Build GRCh38
rs756693851 Chr.2: 218266424-218266424 on Build GRCh38
rs909463675 Chr.2: 218266423-218266423 on Build GRCh38
rs749452251 Chr.2: 218266180-218266180 on Build GRCh38
rs762549283 Chr.2: 218266097-218266097 on Build GRCh38
rs1049677834 Chr.2: 218266139-218266139 on Build GRCh38
rs757503345 Chr.2: 218266399-218266399 on Build GRCh38
rs878986494 Chr.2: 218266441-218266441 on Build GRCh38
rs746227810 Chr.2: 218266048-218266048 on Build GRCh38
rs768740919 Chr.2: 218266060-218266060 on Build GRCh38
rs769543311 Chr.2: 218266196-218266196 on Build GRCh38
rs2305835 Chr.2: 218266079-218266079 on Build GRCh38
rs1052427573 Chr.2: 218266237-218266237 on Build GRCh38
rs535782371 Chr.2: 218266429-218266429 on Build GRCh38
rs956531007 Chr.2: 218266338-218266338 on Build GRCh38
rs777174790 Chr.2: 218266006-218266006 on Build GRCh38
rs966133222 Chr.2: 218266395-218266395 on Build GRCh38
rs202243854 Chr.2: 218266408-218266408 on Build GRCh38
rs774297760 Chr.2: 218266065-218266065 on Build GRCh38
rs933390734 Chr.2: 218266265-218266265 on Build GRCh38
rs923294148 Chr.2: 218266262-218266262 on Build GRCh38
rs781750286 Chr.2: 218266047-218266047 on Build GRCh38
rs551476250 Chr.2: 218266356-218266356 on Build GRCh38
rs752068817 Chr.2: 218266027-218266027 on Build GRCh38
rs772234812 Chr.2: 218266189-218266189 on Build GRCh38
rs200612755 Chr.2: 218266389-218266389 on Build GRCh38
rs148400911 Chr.2: 218266182-218266182 on Build GRCh38
rs551028094 Chr.2: 218266214-218266214 on Build GRCh38
rs987973657 Chr.2: 218266352-218266352 on Build GRCh38
rs533317601 Chr.2: 218266337-218266337 on Build GRCh38
rs376390666 Chr.2: 218265997-218265997 on Build GRCh38
rs969691774 Chr.2: 218266302-218266302 on Build GRCh38
rs770762630 Chr.2: 218266075-218266075 on Build GRCh38
rs749542172 Chr.2: 218266054-218266054 on Build GRCh38
rs751082045 Chr.2: 218266351-218266351 on Build GRCh38
rs765066264 Chr.2: 218266081-218266081 on Build GRCh38
rs751047477 Chr.2: 218266153-218266153 on Build GRCh38
rs993737328 Chr.2: 218266391-218266391 on Build GRCh38
rs779587714 Chr.2: 218266218-218266218 on Build GRCh38
rs748147108 Chr.2: 218266016-218266017 on Build GRCh38
rs778974024 Chr.2: 218266433-218266433 on Build GRCh38
rs748174891 Chr.2: 218266186-218266186 on Build GRCh38

View Full Product Details