The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014681.5 | NP_055496.2 | 5 - 6 | 1723 | 59 |
XM_005259443.3 | XP_005259500.1 | 5 - 6 | 2234 | 59 | |
XM_011527550.2 | XP_011525852.1 | 5 - 6 | 2358 | 59 | |
XM_011527551.2 | XP_011525853.1 | 5 - 6 | 1672 | 59 | |
GenBank mRNA | BQ708034.1 | - | 3 - 4 | 538 | 59 |
CF125648.1 | - | 2 - 3 | 359 | 59 | |
D50924.1 | - | 3 - 4 | 1405 | 59 |