Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219239477-219239985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLB1L
Entrez Gene ID
79411
Gene Name
galactosidase beta 1 like
Gene Aliases
-
UniGene
Hs.181173
Gene Chromosome Location
Chr.2: 219236598-219245429 on Build GRCh38
Transcripts
NM_001286423.1 NM_001286427.1 NM_024506.4 XM_011511813.2 XM_017004895.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3044732 c.902T>C p.M301T Chr.2: 219239561-219239561 on Build GRCh38
1405600 c.839G>A p.R280Q Chr.2: 219239624-219239624 on Build GRCh38
442343 c.790G>A p.E264K Chr.2: 219239673-219239673 on Build GRCh38
1016656 c.825G>T p.Q275H Chr.2: 219239638-219239638 on Build GRCh38
1016657 c.721+9C>T p.? Chr.2: 219239911-219239911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763648085 Chr.2: 219239606-219239606 on Build GRCh38
rs150577551 Chr.2: 219239624-219239624 on Build GRCh38
rs985912258 Chr.2: 219239846-219239846 on Build GRCh38
rs772765930 Chr.2: 219239832-219239832 on Build GRCh38
rs775337435 Chr.2: 219239686-219239686 on Build GRCh38
rs779750472 Chr.2: 219239809-219239809 on Build GRCh38
rs768390059 Chr.2: 219239814-219239814 on Build GRCh38
rs749059558 Chr.2: 219239811-219239811 on Build GRCh38
rs199578439 Chr.2: 219239610-219239610 on Build GRCh38
rs770300817 Chr.2: 219239908-219239908 on Build GRCh38
rs751209809 Chr.2: 219239827-219239827 on Build GRCh38
rs761332404 Chr.2: 219239734-219239734 on Build GRCh38
rs906545054 Chr.2: 219239856-219239856 on Build GRCh38
rs749004476 Chr.2: 219239697-219239697 on Build GRCh38
rs779202174 Chr.2: 219239648-219239648 on Build GRCh38
rs994001796 Chr.2: 219239848-219239848 on Build GRCh38
rs879438522 Chr.2: 219239863-219239863 on Build GRCh38
rs761155299 Chr.2: 219239837-219239837 on Build GRCh38
rs143841404 Chr.2: 219239674-219239674 on Build GRCh38
rs1012977310 Chr.2: 219239732-219239732 on Build GRCh38
rs113157998 Chr.2: 219239866-219239866 on Build GRCh38
rs957900970 Chr.2: 219239647-219239647 on Build GRCh38
rs926501574 Chr.2: 219239643-219239643 on Build GRCh38
rs73991428 Chr.2: 219239876-219239876 on Build GRCh38
rs373530917 Chr.2: 219239788-219239788 on Build GRCh38
rs759737823 Chr.2: 219239671-219239671 on Build GRCh38
rs187487533 Chr.2: 219239759-219239759 on Build GRCh38
rs541235357 Chr.2: 219239952-219239952 on Build GRCh38
rs764142790 Chr.2: 219239868-219239868 on Build GRCh38
rs199831758 Chr.2: 219239961-219239961 on Build GRCh38
rs754227381 Chr.2: 219239782-219239782 on Build GRCh38
rs568426012 Chr.2: 219239825-219239825 on Build GRCh38
rs779077511 Chr.2: 219239536-219239536 on Build GRCh38
rs766712093 Chr.2: 219239781-219239781 on Build GRCh38
rs746508436 Chr.2: 219239903-219239903 on Build GRCh38
rs143823898 Chr.2: 219239800-219239800 on Build GRCh38
rs146996880 Chr.2: 219239921-219239921 on Build GRCh38
rs754021201 Chr.2: 219239637-219239637 on Build GRCh38
rs200552871 Chr.2: 219239574-219239574 on Build GRCh38
rs750891160 Chr.2: 219239625-219239625 on Build GRCh38
rs757316142 Chr.2: 219239663-219239663 on Build GRCh38
rs552422506 Chr.2: 219239629-219239629 on Build GRCh38
rs768263440 Chr.2: 219239705-219239705 on Build GRCh38
rs751625628 Chr.2: 219239869-219239869 on Build GRCh38
rs192758062 Chr.2: 219239871-219239871 on Build GRCh38
rs534291035 Chr.2: 219239756-219239756 on Build GRCh38
rs761058743 Chr.2: 219239759-219239760 on Build GRCh38
rs756072424 Chr.2: 219239808-219239808 on Build GRCh38
rs770990866 Chr.2: 219239554-219239554 on Build GRCh38
rs980663643 Chr.2: 219239631-219239631 on Build GRCh38
rs370495901 Chr.2: 219239906-219239906 on Build GRCh38
rs144186863 Chr.2: 219239627-219239627 on Build GRCh38
rs370083958 Chr.2: 219239789-219239789 on Build GRCh38
rs765661804 Chr.2: 219239843-219239843 on Build GRCh38
rs775006335 Chr.2: 219239600-219239600 on Build GRCh38
rs745630727 Chr.2: 219239589-219239589 on Build GRCh38
rs775798513 Chr.2: 219239695-219239696 on Build GRCh38
rs932267470 Chr.2: 219239810-219239810 on Build GRCh38
rs986282379 Chr.2: 219239542-219239542 on Build GRCh38
rs755239333 Chr.2: 219239646-219239646 on Build GRCh38
rs781017676 Chr.2: 219239668-219239668 on Build GRCh38
rs747002150 Chr.2: 219239549-219239549 on Build GRCh38
rs771439705 Chr.2: 219239838-219239838 on Build GRCh38
rs966537660 Chr.2: 219239919-219239919 on Build GRCh38
rs1044497532 Chr.2: 219239953-219239953 on Build GRCh38
rs764386196 Chr.2: 219239810-219239811 on Build GRCh38
rs149500487 Chr.2: 219239964-219239964 on Build GRCh38
rs767013047 Chr.2: 219239741-219239741 on Build GRCh38
rs373436592 Chr.2: 219239595-219239595 on Build GRCh38
rs550201712 Chr.2: 219239601-219239601 on Build GRCh38
rs755114581 Chr.2: 219239521-219239521 on Build GRCh38
rs769082524 Chr.2: 219239561-219239561 on Build GRCh38
rs770336668 Chr.2: 219239924-219239924 on Build GRCh38
rs748210483 Chr.2: 219239653-219239653 on Build GRCh38
rs932337831 Chr.2: 219239956-219239956 on Build GRCh38
rs773898356 Chr.2: 219239729-219239729 on Build GRCh38
rs201325778 Chr.2: 219239801-219239801 on Build GRCh38
rs771752415 Chr.2: 219239896-219239896 on Build GRCh38
rs929288853 Chr.2: 219239658-219239658 on Build GRCh38
rs945138491 Chr.2: 219239780-219239780 on Build GRCh38
rs778620160 Chr.2: 219239888-219239888 on Build GRCh38
rs754313026 Chr.2: 219239890-219239891 on Build GRCh38
rs780062250 Chr.2: 219239875-219239875 on Build GRCh38
rs767772402 Chr.2: 219239694-219239695 on Build GRCh38
rs780160155 Chr.2: 219239517-219239517 on Build GRCh38
rs752922483 Chr.2: 219239793-219239793 on Build GRCh38
rs561207688 Chr.2: 219239520-219239520 on Build GRCh38
rs74865497 Chr.2: 219239821-219239821 on Build GRCh38
rs754771308 Chr.2: 219239887-219239887 on Build GRCh38
rs74566954 Chr.2: 219239841-219239841 on Build GRCh38
rs763219544 Chr.2: 219239854-219239854 on Build GRCh38
rs763122303 Chr.2: 219239960-219239960 on Build GRCh38
rs774423137 Chr.2: 219239652-219239665 on Build GRCh38
rs144269036 Chr.2: 219239891-219239891 on Build GRCh38
rs145811809 Chr.2: 219239684-219239684 on Build GRCh38
rs753923117 Chr.2: 219239938-219239938 on Build GRCh38
rs760057694 Chr.2: 219239922-219239922 on Build GRCh38
rs752192966 Chr.2: 219239898-219239898 on Build GRCh38
rs376236263 Chr.2: 219239844-219239844 on Build GRCh38
rs773596639 Chr.2: 219239742-219239742 on Build GRCh38

View Full Product Details