The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014762.3 | NP_055577.1 | 1 - 2 | 360 | 51 |
GenBank mRNA | AB073391.1 | - | 2 - 3 | 235 | 51 |
AF261758.1 | - | 1 - 2 | 330 | 51 | |
AK304421.1 | - | 1 - 2 | 309 | 51 | |
AK316427.1 | - | 1 - 2 | 61 | 51 | |
BC004375.1 | - | 1 - 2 | 266 | 51 | |
BC011669.2 | - | 1 - 2 | 293 | 51 | |
BP200916.1 | - | 1 - 2 | 360 | 51 | |
D13643.3 | - | 1 - 2 | 343 | 51 |