Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 227519862-227520372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGFG1
Entrez Gene ID
3267
Gene Name
ArfGAP with FG repeats 1
Gene Aliases
HRB, RAB, RIP
UniGene
Hs.352962
Gene Chromosome Location
Chr.2: 227472169-227561222 on Build GRCh38
Transcripts
NM_001135187.1 NM_001135188.1 NM_001135189.1 NM_004504.4 XM_005246516.3 XM_006712478.3 XM_006712479.3 XM_006712480.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1405980 c.337G>T p.V113L Chr.2: 227520023-227520023 on Build GRCh38
6380881 c.308C>A p.A103E Chr.2: 227519994-227519994 on Build GRCh38
4419077 c.333A>G p.Q111Q Chr.2: 227520019-227520019 on Build GRCh38
5861936 c.314C>T p.P105L Chr.2: 227520000-227520000 on Build GRCh38
5861935 c.266G>A p.C89Y Chr.2: 227519952-227519952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746362567 Chr.2: 227520037-227520037 on Build GRCh38
rs762325021 Chr.2: 227520114-227520114 on Build GRCh38
rs200608192 Chr.2: 227520125-227520125 on Build GRCh38
rs765291061 Chr.2: 227519987-227519987 on Build GRCh38
rs1028440189 Chr.2: 227520132-227520132 on Build GRCh38
rs757662350 Chr.2: 227520027-227520027 on Build GRCh38
rs780300802 Chr.2: 227520230-227520230 on Build GRCh38
rs552566009 Chr.2: 227519927-227519927 on Build GRCh38
rs773194882 Chr.2: 227520102-227520102 on Build GRCh38
rs762917289 Chr.2: 227520110-227520110 on Build GRCh38
rs767494345 Chr.2: 227519953-227519953 on Build GRCh38
rs189343739 Chr.2: 227520229-227520229 on Build GRCh38
rs554314063 Chr.2: 227520218-227520218 on Build GRCh38
rs768439278 Chr.2: 227520259-227520259 on Build GRCh38
rs1019901077 Chr.2: 227520117-227520117 on Build GRCh38
rs752665394 Chr.2: 227519995-227519995 on Build GRCh38
rs967280629 Chr.2: 227520311-227520311 on Build GRCh38
rs761468129 Chr.2: 227519938-227519938 on Build GRCh38
rs781780869 Chr.2: 227520032-227520032 on Build GRCh38
rs113506143 Chr.2: 227519918-227519918 on Build GRCh38
rs774233577 Chr.2: 227519933-227519933 on Build GRCh38
rs748018572 Chr.2: 227520094-227520094 on Build GRCh38
rs771853645 Chr.2: 227520099-227520099 on Build GRCh38
rs950049257 Chr.2: 227520270-227520270 on Build GRCh38
rs1135969 Chr.2: 227520019-227520019 on Build GRCh38
rs952445982 Chr.2: 227520176-227520176 on Build GRCh38
rs554156591 Chr.2: 227520302-227520302 on Build GRCh38
rs1001332685 Chr.2: 227520207-227520207 on Build GRCh38
rs201911317 Chr.2: 227519925-227519925 on Build GRCh38
rs756630641 Chr.2: 227520039-227520039 on Build GRCh38
rs764169526 Chr.2: 227520013-227520013 on Build GRCh38
rs959895214 Chr.2: 227520251-227520251 on Build GRCh38
rs775340289 Chr.2: 227519899-227519899 on Build GRCh38
rs766330564 Chr.2: 227519915-227519916 on Build GRCh38
rs955045744 Chr.2: 227520141-227520141 on Build GRCh38
rs376341887 Chr.2: 227520108-227520108 on Build GRCh38
rs972666096 Chr.2: 227520183-227520183 on Build GRCh38
rs751258123 Chr.2: 227520079-227520079 on Build GRCh38
rs139973556 Chr.2: 227520289-227520289 on Build GRCh38
rs773104819 Chr.2: 227519976-227519976 on Build GRCh38
rs758674049 Chr.2: 227520008-227520008 on Build GRCh38
rs748983488 Chr.2: 227519921-227519921 on Build GRCh38
rs889994242 Chr.2: 227520081-227520081 on Build GRCh38
rs773854548 Chr.2: 227520086-227520086 on Build GRCh38
rs779620457 Chr.2: 227520051-227520051 on Build GRCh38
rs535941459 Chr.2: 227520205-227520205 on Build GRCh38
rs574783550 Chr.2: 227520192-227520192 on Build GRCh38
rs925345547 Chr.2: 227520332-227520332 on Build GRCh38
rs962174903 Chr.2: 227520180-227520180 on Build GRCh38
rs751724424 Chr.2: 227520112-227520112 on Build GRCh38
rs781350818 Chr.2: 227520253-227520253 on Build GRCh38
rs1045680877 Chr.2: 227519943-227519943 on Build GRCh38
rs897427426 Chr.2: 227520159-227520159 on Build GRCh38
rs17852771 Chr.2: 227519956-227519956 on Build GRCh38
rs759682631 Chr.2: 227519982-227519982 on Build GRCh38
rs915874402 Chr.2: 227520237-227520237 on Build GRCh38
rs777960267 Chr.2: 227520106-227520108 on Build GRCh38
rs994390732 Chr.2: 227520162-227520162 on Build GRCh38
rs566385884 Chr.2: 227519926-227519926 on Build GRCh38
rs979046924 Chr.2: 227520329-227520329 on Build GRCh38
rs758590740 Chr.2: 227520175-227520175 on Build GRCh38
rs370784434 Chr.2: 227520111-227520111 on Build GRCh38
rs751398868 Chr.2: 227519922-227519923 on Build GRCh38

View Full Product Details