Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 227609725-227610267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf83
Entrez Gene ID
56918
Gene Name
chromosome 2 open reading frame 83
Gene Aliases
-
UniGene
Hs.283092
Gene Chromosome Location
Chr.2: 227610090-227633320 on Build GRCh38
Transcripts
NM_001162483.1 NM_020161.3

SNP Detail

SNP ID Chromosome Location
rs868098841 Chr.2: 227609966-227609966 on Build GRCh38
rs10648670 Chr.2: 227610055-227610056 on Build GRCh38
rs77664074 Chr.2: 227610238-227610238 on Build GRCh38
rs112228641 Chr.2: 227609907-227609907 on Build GRCh38
rs11689897 Chr.2: 227610076-227610076 on Build GRCh38
rs991572143 Chr.2: 227610126-227610126 on Build GRCh38
rs993591038 Chr.2: 227610084-227610084 on Build GRCh38
rs985550045 Chr.2: 227609775-227609775 on Build GRCh38
rs571307481 Chr.2: 227610037-227610037 on Build GRCh38
rs1041998845 Chr.2: 227609965-227609965 on Build GRCh38
rs3791208 Chr.2: 227609976-227609976 on Build GRCh38
rs1018685601 Chr.2: 227609782-227609782 on Build GRCh38
rs532196006 Chr.2: 227610111-227610111 on Build GRCh38
rs962155545 Chr.2: 227609779-227609779 on Build GRCh38
rs367552982 Chr.2: 227609819-227609819 on Build GRCh38
rs559678058 Chr.2: 227610047-227610048 on Build GRCh38
rs776449408 Chr.2: 227609984-227609984 on Build GRCh38
rs958118916 Chr.2: 227609774-227609774 on Build GRCh38
rs924454251 Chr.2: 227609927-227609927 on Build GRCh38
rs1027772483 Chr.2: 227610110-227610110 on Build GRCh38
rs1025865602 Chr.2: 227610138-227610138 on Build GRCh38
rs1004666585 Chr.2: 227610038-227610038 on Build GRCh38
rs965734905 Chr.2: 227610048-227610048 on Build GRCh38
rs546605611 Chr.2: 227610019-227610019 on Build GRCh38
rs758980814 Chr.2: 227610237-227610237 on Build GRCh38
rs772825840 Chr.2: 227610007-227610007 on Build GRCh38
rs949767820 Chr.2: 227609950-227609950 on Build GRCh38
rs542873310 Chr.2: 227609942-227609942 on Build GRCh38
rs376044099 Chr.2: 227610075-227610075 on Build GRCh38
rs888984020 Chr.2: 227610029-227610029 on Build GRCh38
rs981371375 Chr.2: 227609918-227609918 on Build GRCh38
rs1032804981 Chr.2: 227610011-227610011 on Build GRCh38
rs981786123 Chr.2: 227610188-227610188 on Build GRCh38
rs762650321 Chr.2: 227610135-227610135 on Build GRCh38
rs769614264 Chr.2: 227609999-227609999 on Build GRCh38
rs972141018 Chr.2: 227609784-227609784 on Build GRCh38
rs145838330 Chr.2: 227609820-227609820 on Build GRCh38
rs1009577133 Chr.2: 227609987-227609987 on Build GRCh38
rs924485272 Chr.2: 227610226-227610226 on Build GRCh38
rs541628768 Chr.2: 227609911-227609911 on Build GRCh38
rs930099684 Chr.2: 227609896-227609896 on Build GRCh38
rs762130084 Chr.2: 227610168-227610168 on Build GRCh38
rs952053937 Chr.2: 227609850-227609850 on Build GRCh38
rs34337164 Chr.2: 227610240-227610240 on Build GRCh38
rs773844698 Chr.2: 227610221-227610221 on Build GRCh38
rs747930973 Chr.2: 227609985-227609985 on Build GRCh38
rs561050767 Chr.2: 227609945-227609945 on Build GRCh38
rs2176191 Chr.2: 227610157-227610157 on Build GRCh38
rs1048964376 Chr.2: 227609813-227609813 on Build GRCh38
rs1055581473 Chr.2: 227609919-227609919 on Build GRCh38
rs889403726 Chr.2: 227609768-227609768 on Build GRCh38
rs749213093 Chr.2: 227609758-227609758 on Build GRCh38
rs918863845 Chr.2: 227609840-227609840 on Build GRCh38
rs890782279 Chr.2: 227609977-227609978 on Build GRCh38
rs937069048 Chr.2: 227609912-227609912 on Build GRCh38
rs182573292 Chr.2: 227609861-227609861 on Build GRCh38
rs1023416897 Chr.2: 227610139-227610139 on Build GRCh38
rs1054209135 Chr.2: 227609959-227609959 on Build GRCh38
rs999946607 Chr.2: 227609977-227609977 on Build GRCh38
rs1007371298 Chr.2: 227610030-227610030 on Build GRCh38

View Full Product Details