Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MELK
Entrez Gene ID
9833
Gene Name
maternal embryonic leucine zipper kinase
Gene Aliases
HPK38
Chromosome Location
Chr.9: 36572862 - 36677683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256685.1 NP_001243614.1 5 - 6 593 91
NM_001256689.1 NP_001243618.1 4 - 5 507 91
NM_001256690.1 NP_001243619.1 3 - 4 390 91
NM_001256693.1 NP_001243622.1 3 - 4 411 91
NM_014791.3 NP_055606.1 5 - 6 593 91
NR_046337.1 4 - 5 507 91
XM_011518076.2 XP_011516378.1 5 - 6 772 91
XM_011518077.1 XP_011516379.1 6 - 7 668 91
XM_011518078.2 XP_011516380.1 6 - 7 855 91
XM_011518079.1 XP_011516381.1 5 - 6 545 91
XM_011518080.1 XP_011516382.1 5 - 6 560 91
XM_011518081.2 XP_011516383.1 5 - 6 578 91
XM_011518082.2 XP_011516384.1 4 - 5 686 91
XM_011518083.2 XP_011516385.1 5 - 6 769 91
XM_011518084.2 XP_011516386.1 4 - 5 381 91
XM_011518085.1 XP_011516387.1 3 - 4 377 91
XM_011518086.2 XP_011516388.1 5 - 6 560 91
GenBank mRNA AB183427.1 - 4 - 5 458 91
AB183428.1 - 3 - 4 343 91
AK293447.1 - 3 - 4 347 91
AK296820.1 - 4 - 5 506 91
AK299164.1 - 5 - 6 545 91
AK300821.1 - 4 - 5 461 91
AK301131.1 - 3 - 4 343 91
AK309470.1 - 3 - 4 368 91
BC014039.2 - 5 - 6 547 91
D79997.1 - 5 - 6 579 91

View Full Product Details