Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232409065-232409563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALPPL2
Entrez Gene ID
251
Gene Name
alkaline phosphatase, placental like 2
Gene Aliases
ALPG, ALPPL, GCAP
UniGene
Hs.333509
Gene Chromosome Location
Chr.2: 232406842-232410714 on Build GRCh38
Transcripts
NM_031313.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6050603 c.1237G>A p.G413R Chr.2: 232409385-232409385 on Build GRCh38
6375427 c.1247C>T p.P416L Chr.2: 232409395-232409395 on Build GRCh38
477077 c.1267G>T p.G423C Chr.2: 232409415-232409415 on Build GRCh38
2154687 c.1266C>T p.D422D Chr.2: 232409414-232409414 on Build GRCh38
6192629 c.1253A>G p.Y418C Chr.2: 232409401-232409401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780238836 Chr.2: 232409397-232409397 on Build GRCh38
rs1017377487 Chr.2: 232409169-232409169 on Build GRCh38
rs544925049 Chr.2: 232409364-232409364 on Build GRCh38
rs779069758 Chr.2: 232409415-232409429 on Build GRCh38
rs746873216 Chr.2: 232409422-232409422 on Build GRCh38
rs573331017 Chr.2: 232409235-232409235 on Build GRCh38
rs748973186 Chr.2: 232409409-232409409 on Build GRCh38
rs751635156 Chr.2: 232409371-232409371 on Build GRCh38
rs758770108 Chr.2: 232409497-232409498 on Build GRCh38
rs545566993 Chr.2: 232409236-232409236 on Build GRCh38
rs201572907 Chr.2: 232409353-232409353 on Build GRCh38
rs2091555 Chr.2: 232409459-232409459 on Build GRCh38
rs536268368 Chr.2: 232409204-232409204 on Build GRCh38
rs763229287 Chr.2: 232409354-232409354 on Build GRCh38
rs565969893 Chr.2: 232409465-232409467 on Build GRCh38
rs987306306 Chr.2: 232409362-232409362 on Build GRCh38
rs770792158 Chr.2: 232409425-232409425 on Build GRCh38
rs755331930 Chr.2: 232409454-232409454 on Build GRCh38
rs762084011 Chr.2: 232409523-232409523 on Build GRCh38
rs764472830 Chr.2: 232409430-232409430 on Build GRCh38
rs551153828 Chr.2: 232409447-232409447 on Build GRCh38
rs372779909 Chr.2: 232409338-232409338 on Build GRCh38
rs773251437 Chr.2: 232409419-232409419 on Build GRCh38
rs772054347 Chr.2: 232409290-232409290 on Build GRCh38
rs776621008 Chr.2: 232409483-232409483 on Build GRCh38
rs763284496 Chr.2: 232409428-232409428 on Build GRCh38
rs756509077 Chr.2: 232409441-232409441 on Build GRCh38
rs563231782 Chr.2: 232409384-232409384 on Build GRCh38
rs188679065 Chr.2: 232409222-232409222 on Build GRCh38
rs192580193 Chr.2: 232409508-232409508 on Build GRCh38
rs757404029 Chr.2: 232409375-232409375 on Build GRCh38
rs747777494 Chr.2: 232409457-232409457 on Build GRCh38
rs745728114 Chr.2: 232409484-232409484 on Build GRCh38
rs766067546 Chr.2: 232409343-232409343 on Build GRCh38
rs571157591 Chr.2: 232409456-232409456 on Build GRCh38
rs567230735 Chr.2: 232409192-232409192 on Build GRCh38
rs754566560 Chr.2: 232409411-232409411 on Build GRCh38
rs994552098 Chr.2: 232409196-232409196 on Build GRCh38
rs964492365 Chr.2: 232409381-232409381 on Build GRCh38
rs760788057 Chr.2: 232409532-232409532 on Build GRCh38
rs774732852 Chr.2: 232409498-232409498 on Build GRCh38
rs145179673 Chr.2: 232409415-232409415 on Build GRCh38
rs770719668 Chr.2: 232409481-232409481 on Build GRCh38
rs769686397 Chr.2: 232409492-232409492 on Build GRCh38
rs200776172 Chr.2: 232409329-232409329 on Build GRCh38
rs376573973 Chr.2: 232409499-232409499 on Build GRCh38
rs369960059 Chr.2: 232409271-232409271 on Build GRCh38
rs2091556 Chr.2: 232409524-232409524 on Build GRCh38
rs142455461 Chr.2: 232409418-232409418 on Build GRCh38
rs202082280 Chr.2: 232409177-232409177 on Build GRCh38
rs371606354 Chr.2: 232409231-232409231 on Build GRCh38
rs528210448 Chr.2: 232409436-232409436 on Build GRCh38
rs756520748 Chr.2: 232409385-232409385 on Build GRCh38
rs754257668 Chr.2: 232409452-232409452 on Build GRCh38
rs761751397 Chr.2: 232409136-232409136 on Build GRCh38
rs757352837 Chr.2: 232409320-232409321 on Build GRCh38
rs376010839 Chr.2: 232409256-232409256 on Build GRCh38
rs2090543 Chr.2: 232409435-232409435 on Build GRCh38
rs536673511 Chr.2: 232409525-232409525 on Build GRCh38
rs536903121 Chr.2: 232409458-232409458 on Build GRCh38
rs565518188 Chr.2: 232409277-232409277 on Build GRCh38
rs776225258 Chr.2: 232409426-232409426 on Build GRCh38
rs780342847 Chr.2: 232409523-232409524 on Build GRCh38
rs746972812 Chr.2: 232409462-232409462 on Build GRCh38

View Full Product Details