Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL21
Entrez Gene ID
9903
Gene Name
kelch like family member 21
Gene Aliases
-
Chromosome Location
Chr.1: 6590724 - 6602898 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324309.1 NP_001311238.1 1 - 2 1106 70
NM_014851.3 NP_055666.2 1 - 2 1106 70
GenBank mRNA AB007938.1 - 1 - 2 1209 70
AK075305.1 - 1 - 2 1077 70
AK090472.1 - 1 - 2 875 70
BC064980.1 - 1 - 2 1059 70
BC091648.1 - 1 - 2 1059 70

View Full Product Details