The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014918.4 | NP_055733.2 | 1 - 2 | 796 | 77 |
XM_011521364.2 | XP_011519666.1 | 1 - 2 | 335 | 77 | |
GenBank mRNA | AB023207.1 | - | 1 - 2 | 814 | 77 |
AB071402.1 | - | 1 - 2 | 334 | 77 | |
AY358529.1 | - | 1 - 2 | 808 | 77 | |
BC046247.1 | - | 1 - 2 | 803 | 77 |