The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136193.1 | NP_001129665.1 | 2 - 3 | 1185 | 70 |
NM_001136194.1 | NP_001129666.1 | 2 - 3 | 1200 | 70 | |
NM_014929.3 | NP_055744.2 | 2 - 3 | 1092 | 70 | |
GenBank mRNA | AB023188.1 | - | 1 - 2 | 833 | 70 |
AF223470.1 | - | 2 - 3 | 959 | 70 | |
AK001516.1 | - | 2 - 3 | 872 | 70 | |
AK021557.1 | - | 2 - 3 | 986 | 70 | |
AL833877.1 | - | 2 - 3 | 866 | 70 | |
BC001544.1 | - | 1 - 2 | 832 | 70 |