The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015024.4 | NP_055839.3 | 8 - 9 | 937 | 62 |
XM_017013244.1 | XP_016868733.1 | 8 - 9 | 1191 | 62 | |
XM_017013245.1 | XP_016868734.1 | 8 - 9 | 958 | 62 | |
XM_017013246.1 | XP_016868735.1 | 8 - 9 | 1160 | 62 | |
XM_017013247.1 | XP_016868736.1 | 8 - 9 | 1351 | 62 | |
GenBank mRNA | AF064729.1 | - | 8 - 9 | 847 | 62 |
AK291472.1 | - | 8 - 9 | 852 | 62 | |
BC030785.1 | - | 8 - 9 | 936 | 62 |