The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015227.4 | NP_056042.1 | 4 - 5 | 666 | 61 |
NM_133635.4 | NP_598368.2 | 4 - 5 | 666 | 61 | |
NR_004858.1 | 4 - 5 | 666 | 61 | ||
XM_006723981.3 | XP_006724044.1 | 5 - 6 | 739 | 61 | |
XM_011529504.2 | XP_011527806.1 | 6 - 7 | 875 | 61 | |
XM_011529505.2 | XP_011527807.1 | 6 - 7 | 876 | 61 | |
XM_011529506.2 | XP_011527808.1 | 5 - 6 | 814 | 61 | |
XM_011529507.2 | XP_011527809.1 | 5 - 6 | 710 | 61 | |
XM_011529508.2 | XP_011527810.1 | 6 - 7 | 876 | 61 | |
XM_011529509.2 | XP_011527811.1 | 6 - 7 | 877 | 61 | |
XM_011529510.2 | XP_011527812.1 | 6 - 7 | 784 | 61 | |
XM_011529511.2 | XP_011527813.1 | 6 - 7 | 816 | 61 | |
XM_017028306.1 | XP_016883795.1 | 4 - 5 | 649 | 61 | |
XM_017028309.1 | XP_016883798.1 | 2 - 3 | 72 | 61 | |
GenBank mRNA | AB023175.1 | - | 4 - 5 | 655 | 61 |
AJ302079.2 | - | 4 - 5 | 655 | 61 | |
AJ302080.2 | - | 4 - 5 | 655 | 61 | |
AJ575591.1 | - | 4 - 5 | 650 | 61 | |
AL110285.1 | - | 4 - 5 | 652 | 61 | |
AY066015.2 | - | 4 - 5 | 655 | 61 | |
BC064623.1 | - | 4 - 5 | 648 | 61 |