Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
R3HDM1
Entrez Gene ID
23518
Gene Name
R3H domain containing 1
Gene Aliases
R3HDM
Chromosome Location
Chr.2: 135531429 - 135725270 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282798.1 NP_001269727.1 17 - 18 2361 65
NM_001282799.1 NP_001269728.1 14 - 15 1969 65
NM_001282800.1 NP_001269729.1 14 - 15 2018 65
NM_015361.3 NP_056176.2 17 - 18 2353 65
XM_005263626.3 XP_005263683.1 17 - 18 2159 65
XM_005263627.3 XP_005263684.1 17 - 18 2156 65
XM_005263628.3 XP_005263685.1 16 - 17 2090 65
XM_005263629.3 XP_005263686.1 16 - 17 2087 65
XM_006712389.2 XP_006712452.1 17 - 18 2345 65
XM_011510870.1 XP_011509172.1 18 - 19 2447 65
XM_011510871.1 XP_011509173.1 18 - 19 2463 65
XM_011510872.2 XP_011509174.1 17 - 18 2405 65
XM_011510873.2 XP_011509175.1 18 - 19 2399 65
XM_011510874.2 XP_011509176.1 16 - 17 2315 65
XM_011510875.2 XP_011509177.1 16 - 17 2331 65
XM_011510876.2 XP_011509178.1 18 - 19 2261 65
XM_011510877.2 XP_011509179.1 18 - 19 2213 65
XM_011510878.2 XP_011509180.1 17 - 18 2192 65
XM_011510879.2 XP_011509181.1 17 - 18 2144 65
XM_011510881.1 XP_011509183.1 18 - 19 2447 65
XM_017003718.1 XP_016859207.1 17 - 18 2358 65
XM_017003719.1 XP_016859208.1 16 - 17 2337 65
XM_017003720.1 XP_016859209.1 17 - 18 2310 65
XM_017003721.1 XP_016859210.1 18 - 19 2277 65
XM_017003722.1 XP_016859211.1 15 - 16 2213 65
XM_017003723.1 XP_016859212.1 15 - 16 2229 65
XM_017003724.1 XP_016859213.1 15 - 16 2048 65
XM_017003725.1 XP_016859214.1 19 - 20 2539 65
XM_017003726.1 XP_016859215.1 19 - 20 2609 65
XM_017003727.1 XP_016859216.1 18 - 19 2567 65
XM_017003728.1 XP_016859217.1 16 - 17 2042 65
XM_017003729.1 XP_016859218.1 17 - 18 2384 65
XM_017003730.1 XP_016859219.1 15 - 16 2128 65
XM_017003731.1 XP_016859220.1 18 - 19 2458 65
XM_017003732.1 XP_016859221.1 18 - 19 2433 65
XM_017003733.1 XP_016859222.1 19 - 20 2582 65
XM_017003734.1 XP_016859223.1 20 - 21 2742 65
XM_017003735.1 XP_016859224.1 17 - 18 2340 65
XM_017003736.1 XP_016859225.1 16 - 17 2170 65
GenBank mRNA AK290015.1 - 17 - 18 2247 65
AK297000.1 - 9 - 10 1139 65
BC041093.1 - 17 - 18 2295 65
D21852.1 - 14 - 15 1923 65

View Full Product Details