Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LY96
Entrez Gene ID
23643
Gene Name
lymphocyte antigen 96
Gene Aliases
ESOP-1, MD-2, MD2, ly-96
Chromosome Location
Chr.8: 73991275 - 74099807 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015364.4 NP_056179.3 2 - 3 316 116
GenBank mRNA AB018549.1 - 2 - 3 327 116
AB446498.1 - 2 - 3 202 116
AF168121.1 - 2 - 3 223 116
BC020690.1 - 2 - 3 226 116
BG435728.1 - 2 - 3 317 115

View Full Product Details