The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271874.1 | NP_001258803.1 | 2 - 3 | 893 | 93 |
NM_015511.4 | NP_056326.2 | 2 - 3 | 1172 | 93 | |
XM_006723770.3 | XP_006723833.1 | 3 - 4 | 1609 | 93 | |
XM_011528762.2 | XP_011527064.1 | 3 - 4 | 983 | 93 | |
XM_011528763.2 | XP_011527065.1 | 3 - 4 | 1252 | 93 | |
GenBank mRNA | AF113672.1 | - | 1 - 2 | 77 | 93 |
AF132957.1 | - | 1 - 2 | 813 | 93 | |
AL117419.1 | - | 2 - 3 | 833 | 93 | |
BC001751.1 | - | 1 - 2 | 820 | 93 | |
BC019311.1 | - | 2 - 3 | 1064 | 93 |