Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 70259475-70259996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD13C
Entrez Gene ID
81573
Gene Name
ankyrin repeat domain 13C
Gene Aliases
dJ677H15.3
UniGene
Hs.744989
Gene Chromosome Location
Chr.1: 70259002-70354977 on Build GRCh38
Transcripts
NM_030816.4 XM_005271234.3 XM_017002413.1 XM_017002416.1

SNP Detail

SNP ID Chromosome Location
rs1020169667 Chr.1: 70259872-70259872 on Build GRCh38
rs909295418 Chr.1: 70259724-70259724 on Build GRCh38
rs999251251 Chr.1: 70259887-70259887 on Build GRCh38
rs928314220 Chr.1: 70259640-70259643 on Build GRCh38
rs748281602 Chr.1: 70259653-70259653 on Build GRCh38
rs903586915 Chr.1: 70259852-70259852 on Build GRCh38
rs149035868 Chr.1: 70259570-70259570 on Build GRCh38
rs944314320 Chr.1: 70259778-70259778 on Build GRCh38
rs1022014631 Chr.1: 70259510-70259510 on Build GRCh38
rs903084191 Chr.1: 70259927-70259927 on Build GRCh38
rs540339741 Chr.1: 70259888-70259888 on Build GRCh38
rs918393917 Chr.1: 70259781-70259781 on Build GRCh38
rs760830356 Chr.1: 70259816-70259816 on Build GRCh38
rs942335665 Chr.1: 70259827-70259827 on Build GRCh38
rs1006569931 Chr.1: 70259509-70259509 on Build GRCh38
rs1047251955 Chr.1: 70259775-70259775 on Build GRCh38
rs779239289 Chr.1: 70259586-70259586 on Build GRCh38
rs992790153 Chr.1: 70259589-70259589 on Build GRCh38
rs967823777 Chr.1: 70259515-70259515 on Build GRCh38
rs933853847 Chr.1: 70259789-70259789 on Build GRCh38
rs999681405 Chr.1: 70259556-70259556 on Build GRCh38
rs552610332 Chr.1: 70259817-70259817 on Build GRCh38
rs920065725 Chr.1: 70259728-70259728 on Build GRCh38
rs957211920 Chr.1: 70259538-70259538 on Build GRCh38
rs771620518 Chr.1: 70259830-70259830 on Build GRCh38
rs981127828 Chr.1: 70259618-70259618 on Build GRCh38
rs899846046 Chr.1: 70259787-70259789 on Build GRCh38
rs958453405 Chr.1: 70259709-70259709 on Build GRCh38
rs181070333 Chr.1: 70259772-70259772 on Build GRCh38
rs1054284536 Chr.1: 70259801-70259801 on Build GRCh38
rs969319330 Chr.1: 70259659-70259659 on Build GRCh38
rs1020221659 Chr.1: 70259574-70259574 on Build GRCh38
rs577277468 Chr.1: 70259777-70259777 on Build GRCh38
rs143009859 Chr.1: 70259793-70259793 on Build GRCh38
rs984792489 Chr.1: 70259715-70259715 on Build GRCh38
rs916845017 Chr.1: 70259639-70259639 on Build GRCh38
rs780074296 Chr.1: 70259696-70259696 on Build GRCh38
rs578162604 Chr.1: 70259813-70259813 on Build GRCh38
rs772303063 Chr.1: 70259697-70259697 on Build GRCh38
rs375461765 Chr.1: 70259974-70259974 on Build GRCh38

View Full Product Details