Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 70260559-70261096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD13C
Entrez Gene ID
81573
Gene Name
ankyrin repeat domain 13C
Gene Aliases
dJ677H15.3
UniGene
Hs.744989
Gene Chromosome Location
Chr.1: 70259002-70354977 on Build GRCh38
Transcripts
NM_030816.4 XM_005271234.3 XM_005271235.3 XM_006710929.3 XM_017002413.1 XM_017002414.1 XM_017002415.1 XM_017002416.1

SNP Detail

SNP ID Chromosome Location
rs750368739 Chr.1: 70260816-70260816 on Build GRCh38
rs138073712 Chr.1: 70260742-70260742 on Build GRCh38
rs989464342 Chr.1: 70260604-70260604 on Build GRCh38
rs139952550 Chr.1: 70260985-70260985 on Build GRCh38
rs559835825 Chr.1: 70261009-70261009 on Build GRCh38
rs1057003201 Chr.1: 70260642-70260642 on Build GRCh38
rs188557539 Chr.1: 70260824-70260824 on Build GRCh38
rs374685278 Chr.1: 70260636-70260636 on Build GRCh38
rs954428122 Chr.1: 70260791-70260791 on Build GRCh38
rs988197040 Chr.1: 70260952-70260952 on Build GRCh38
rs926379164 Chr.1: 70261002-70261002 on Build GRCh38
rs956676219 Chr.1: 70260939-70260939 on Build GRCh38
rs545271291 Chr.1: 70260817-70260817 on Build GRCh38
rs142561130 Chr.1: 70260831-70260831 on Build GRCh38
rs965193476 Chr.1: 70260741-70260741 on Build GRCh38
rs1003881510 Chr.1: 70260721-70260721 on Build GRCh38
rs758757353 Chr.1: 70260644-70260644 on Build GRCh38
rs576358087 Chr.1: 70260764-70260764 on Build GRCh38
rs898116371 Chr.1: 70260759-70260759 on Build GRCh38
rs185923282 Chr.1: 70260928-70260928 on Build GRCh38
rs902984246 Chr.1: 70260626-70260626 on Build GRCh38
rs747130972 Chr.1: 70260695-70260695 on Build GRCh38
rs925180331 Chr.1: 70260880-70260880 on Build GRCh38
rs934986735 Chr.1: 70260603-70260603 on Build GRCh38
rs746311039 Chr.1: 70261046-70261046 on Build GRCh38
rs1057352764 Chr.1: 70260903-70260903 on Build GRCh38
rs530764467 Chr.1: 70260995-70260995 on Build GRCh38
rs146031924 Chr.1: 70260958-70260958 on Build GRCh38
rs532463539 Chr.1: 70260992-70260992 on Build GRCh38
rs1037879508 Chr.1: 70260740-70260740 on Build GRCh38
rs181092313 Chr.1: 70260876-70260876 on Build GRCh38
rs1006354624 Chr.1: 70260698-70260698 on Build GRCh38
rs1015306891 Chr.1: 70260723-70260723 on Build GRCh38
rs79679858 Chr.1: 70260660-70260660 on Build GRCh38
rs917647606 Chr.1: 70260625-70260625 on Build GRCh38
rs1012862046 Chr.1: 70260654-70260654 on Build GRCh38
rs771440958 Chr.1: 70260991-70260991 on Build GRCh38
rs541544995 Chr.1: 70260843-70260843 on Build GRCh38
rs574076722 Chr.1: 70260623-70260623 on Build GRCh38
rs963830739 Chr.1: 70260849-70260849 on Build GRCh38
rs915274503 Chr.1: 70260614-70260614 on Build GRCh38
rs949189522 Chr.1: 70260649-70260649 on Build GRCh38

View Full Product Details