Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 70261766-70262309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD13C
Entrez Gene ID
81573
Gene Name
ankyrin repeat domain 13C
Gene Aliases
dJ677H15.3
UniGene
Hs.744989
Gene Chromosome Location
Chr.1: 70259002-70354977 on Build GRCh38
Transcripts
NM_030816.4 XM_005271234.3 XM_005271235.3 XM_006710929.3 XM_017002413.1 XM_017002414.1 XM_017002415.1 XM_017002416.1

SNP Detail

SNP ID Chromosome Location
rs919003823 Chr.1: 70261888-70261888 on Build GRCh38
rs543507260 Chr.1: 70261838-70261838 on Build GRCh38
rs77435924 Chr.1: 70262269-70262269 on Build GRCh38
rs370331975 Chr.1: 70262191-70262191 on Build GRCh38
rs767160513 Chr.1: 70262035-70262035 on Build GRCh38
rs1009396190 Chr.1: 70262158-70262158 on Build GRCh38
rs933662492 Chr.1: 70261857-70261858 on Build GRCh38
rs1043497924 Chr.1: 70262182-70262182 on Build GRCh38
rs563483517 Chr.1: 70262095-70262095 on Build GRCh38
rs1024082391 Chr.1: 70261979-70261979 on Build GRCh38
rs200510571 Chr.1: 70261858-70261858 on Build GRCh38
rs896766277 Chr.1: 70261973-70261973 on Build GRCh38
rs559429420 Chr.1: 70262223-70262223 on Build GRCh38
rs902699370 Chr.1: 70261941-70261941 on Build GRCh38
rs929486848 Chr.1: 70261977-70261977 on Build GRCh38
rs985130141 Chr.1: 70262131-70262131 on Build GRCh38
rs963027819 Chr.1: 70261851-70261851 on Build GRCh38
rs910901286 Chr.1: 70262147-70262147 on Build GRCh38
rs984853421 Chr.1: 70262039-70262039 on Build GRCh38
rs1057075006 Chr.1: 70262281-70262281 on Build GRCh38
rs912131737 Chr.1: 70262016-70262016 on Build GRCh38
rs1039323854 Chr.1: 70262091-70262091 on Build GRCh38
rs972695230 Chr.1: 70261819-70261819 on Build GRCh38
rs973141817 Chr.1: 70261853-70261853 on Build GRCh38
rs772605231 Chr.1: 70262137-70262137 on Build GRCh38
rs1035287948 Chr.1: 70261956-70261956 on Build GRCh38
rs1046688450 Chr.1: 70262210-70262210 on Build GRCh38
rs754816676 Chr.1: 70262219-70262219 on Build GRCh38
rs79544665 Chr.1: 70261968-70261968 on Build GRCh38
rs574469596 Chr.1: 70261836-70261836 on Build GRCh38
rs922541006 Chr.1: 70262108-70262108 on Build GRCh38
rs532298399 Chr.1: 70262100-70262100 on Build GRCh38
rs946477466 Chr.1: 70261889-70261889 on Build GRCh38
rs988077857 Chr.1: 70261993-70261993 on Build GRCh38
rs78850336 Chr.1: 70262116-70262116 on Build GRCh38
rs906441965 Chr.1: 70262252-70262252 on Build GRCh38
rs1043886404 Chr.1: 70261904-70261904 on Build GRCh38
rs1000145240 Chr.1: 70261951-70261951 on Build GRCh38
rs1016700572 Chr.1: 70262085-70262085 on Build GRCh38
rs1033565141 Chr.1: 70262274-70262274 on Build GRCh38
rs79336526 Chr.1: 70261967-70261967 on Build GRCh38
rs891915125 Chr.1: 70262148-70262148 on Build GRCh38
rs765839290 Chr.1: 70261870-70261870 on Build GRCh38
rs922223001 Chr.1: 70261820-70261820 on Build GRCh38

View Full Product Details