The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015891.2 | NP_056975.1 | 9 - 10 | 1011 | 77 |
GenBank mRNA | AB208903.1 | - | 9 - 10 | 985 | 77 |
AF038392.1 | - | 9 - 10 | 1011 | 77 | |
AF061241.1 | - | 9 - 10 | 984 | 77 | |
AK222721.1 | - | 9 - 10 | 77 | ||
AK295055.1 | - | 9 - 10 | 1003 | 77 | |
AK314601.1 | - | 9 - 10 | 1007 | 77 | |
BC111061.1 | - | 9 - 10 | 2417 | 77 | |
BC117145.1 | - | 9 - 10 | 1008 | 77 | |
BC126114.1 | - | 9 - 10 | 1008 | 77 |