The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015981.3 | NP_057065.2 | 15 - 16 | 1286 | 76 |
NM_171825.2 | NP_741960.1 | 14 - 15 | 1253 | 76 | |
XM_017009898.1 | XP_016865387.1 | 14 - 15 | 1253 | 76 | |
GenBank mRNA | AB023185.1 | - | 14 - 15 | 1186 | 76 |
AF091486.1 | - | 14 - 15 | 1142 | 76 | |
AF145710.1 | - | 14 - 15 | 1044 | 76 | |
AF145711.1 | - | 15 - 16 | 1077 | 76 | |
AK289776.1 | - | 15 - 16 | 1286 | 76 | |
BC040457.1 | - | 14 - 15 | 1168 | 76 |