Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEBP1
Entrez Gene ID
50865
Gene Name
heme binding protein 1
Gene Aliases
HBP, HEBP
Chromosome Location
Chr.12: 12974864 - 13000309 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015987.4 NP_057071.2 3 - 4 599 62
GenBank mRNA AF117615.1 - 3 - 4 482 62
AF167473.1 - 3 - 4 404 62
AK289877.1 - 3 - 4 563 62
BC016277.2 - 3 - 4 434 62
BT007294.1 - 3 - 4 404 62
CF126276.1 - 3 - 4 600 62

View Full Product Details