The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016034.4 | NP_057118.1 | 2 - 3 | 252 | 122 |
NR_051967.1 | 2 - 3 | 252 | 122 | ||
NR_051968.1 | 2 - 3 | 252 | 122 | ||
XM_006717136.3 | XP_006717199.1 | 3 - 4 | 376 | 122 | |
GenBank mRNA | AB055388.1 | - | 2 - 3 | 176 | 122 |
AF151849.1 | - | 2 - 3 | 176 | 122 | |
AL555185.3 | - | 2 - 3 | 177 | 122 | |
BC008017.1 | - | 2 - 3 | 188 | 122 | |
BC013108.2 | - | 2 - 3 | 196 | 122 | |
BI260471.1 | - | 2 - 3 | 251 | 122 | |
BQ723448.1 | - | 2 - 3 | 184 | 122 | |
BX362434.2 | - | 2 - 3 | 246 | 122 |