The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016053.3 | NP_057137.1 | 4 - 5 | 488 | 115 |
XM_011538423.1 | XP_011536725.1 | 4 - 5 | 406 | 115 | |
XM_017019383.1 | XP_016874872.1 | 3 - 4 | 303 | 115 | |
GenBank mRNA | AF151874.1 | - | 4 - 5 | 433 | 115 |
AF155655.1 | - | 4 - 5 | 433 | 115 | |
AK223339.1 | - | 4 - 5 | 424 | 115 | |
AK314970.1 | - | 4 - 5 | 406 | 115 | |
CR457171.1 | - | 4 - 5 | 326 | 115 |