The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282609.1 | NP_001269538.1 | 1 - 2 | 218 | 63 |
NM_016056.3 | NP_057140.2 | 1 - 2 | 218 | 63 | |
GenBank mRNA | AF113127.1 | - | 1 - 2 | 140 | 63 |
AF151877.1 | - | 1 - 2 | 157 | 63 | |
AF161526.1 | - | 1 - 2 | 109 | 63 | |
AF182041.1 | - | 1 - 2 | 145 | 63 | |
AK075069.1 | - | 1 - 2 | 132 | 63 | |
AK225158.1 | - | 1 - 2 | 138 | 63 | |
BC020613.1 | - | 1 - 2 | 121 | 63 | |
BC052273.1 | - | 1 - 2 | 145 | 63 | |
BC070366.1 | - | 1 - 2 | 144 | 63 | |
DC361259.1 | - | 1 - 2 | 219 | 63 |