The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282750.1 | NP_001269679.1 | 18 - 19 | 2115 | 119 |
NM_001282751.1 | NP_001269680.1 | 17 - 18 | 3068 | 119 | |
NM_014283.4 | NP_055098.1 | 19 - 20 | 3228 | 119 | |
NM_016227.3 | NP_057311.3 | 18 - 19 | 3487 | 119 | |
XM_006711374.2 | XP_006711437.1 | 18 - 19 | 3288 | 119 | |
XM_006711375.2 | XP_006711438.1 | 18 - 19 | 3198 | 119 | |
XM_006711376.1 | XP_006711439.1 | 17 - 18 | 3177 | 119 | |
GenBank mRNA | AF097535.1 | - | 18 - 19 | 3487 | 119 |
AJ250075.1 | - | 19 - 20 | 3226 | 119 | |
AK302509.1 | - | 18 - 19 | 2050 | 119 | |
AK302949.1 | - | 19 - 20 | 3228 | 119 | |
AL035291.1 | - | 19 - 20 | 3129 | 119 | |
BC137125.1 | - | 19 - 20 | 3014 | 119 | |
BC144487.1 | - | 17 - 18 | 2854 | 119 |