Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 25009132-25009632 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSS1
Entrez Gene ID
84532
Gene Name
acyl-CoA synthetase short-chain family member 1
Gene Aliases
ACAS2L, ACECS1, AceCS2L
UniGene
Hs.529353
Gene Chromosome Location
Chr.20: 25006230-25058182 on Build GRCh38
Transcripts
NM_001252675.1 NM_001252676.1 NM_001252677.1 NM_032501.3 XM_006723659.1 XM_011529388.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1025305 c.1789G>A p.V597M Chr.20: 25009371-25009371 on Build GRCh38
6255289 c.1514C>A p.P505H Chr.20: 25009283-25009283 on Build GRCh38
477926 c.1860C>T p.I620I Chr.20: 25009300-25009300 on Build GRCh38
1025304 c.1830G>T p.Q610H Chr.20: 25009330-25009330 on Build GRCh38
6592754 c.1527C>A p.A509A Chr.20: 25009384-25009384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750362489 Chr.20: 25009358-25009358 on Build GRCh38
rs1011518731 Chr.20: 25009483-25009483 on Build GRCh38
rs540694076 Chr.20: 25009355-25009355 on Build GRCh38
rs754351729 Chr.20: 25009220-25009220 on Build GRCh38
rs57913893 Chr.20: 25009161-25009161 on Build GRCh38
rs898013724 Chr.20: 25009387-25009387 on Build GRCh38
rs371407895 Chr.20: 25009224-25009224 on Build GRCh38
rs745950162 Chr.20: 25009222-25009224 on Build GRCh38
rs45555143 Chr.20: 25009437-25009437 on Build GRCh38
rs775641619 Chr.20: 25009413-25009414 on Build GRCh38
rs757019216 Chr.20: 25009255-25009255 on Build GRCh38
rs563074507 Chr.20: 25009434-25009434 on Build GRCh38
rs781118192 Chr.20: 25009260-25009260 on Build GRCh38
rs767441837 Chr.20: 25009357-25009357 on Build GRCh38
rs1034106124 Chr.20: 25009446-25009446 on Build GRCh38
rs745832360 Chr.20: 25009264-25009264 on Build GRCh38
rs187934573 Chr.20: 25009428-25009428 on Build GRCh38
rs777428890 Chr.20: 25009246-25009246 on Build GRCh38
rs768420129 Chr.20: 25009285-25009285 on Build GRCh38
rs765327644 Chr.20: 25009229-25009229 on Build GRCh38
rs754826459 Chr.20: 25009187-25009187 on Build GRCh38
rs1019899854 Chr.20: 25009518-25009518 on Build GRCh38
rs1037886334 Chr.20: 25009370-25009370 on Build GRCh38
rs752899053 Chr.20: 25009231-25009231 on Build GRCh38
rs748883343 Chr.20: 25009395-25009395 on Build GRCh38
rs763073440 Chr.20: 25009329-25009329 on Build GRCh38
rs748915766 Chr.20: 25009283-25009283 on Build GRCh38
rs370228936 Chr.20: 25009514-25009514 on Build GRCh38
rs371706673 Chr.20: 25009423-25009423 on Build GRCh38
rs779706217 Chr.20: 25009378-25009378 on Build GRCh38
rs7347286 Chr.20: 25009223-25009223 on Build GRCh38
rs977286618 Chr.20: 25009589-25009589 on Build GRCh38
rs575005733 Chr.20: 25009322-25009322 on Build GRCh38
rs766965939 Chr.20: 25009219-25009219 on Build GRCh38
rs965698380 Chr.20: 25009579-25009579 on Build GRCh38
rs539350442 Chr.20: 25009174-25009174 on Build GRCh38
rs542081322 Chr.20: 25009591-25009591 on Build GRCh38
rs778623957 Chr.20: 25009402-25009402 on Build GRCh38
rs141779004 Chr.20: 25009404-25009404 on Build GRCh38
rs756160477 Chr.20: 25009373-25009373 on Build GRCh38
rs1021576735 Chr.20: 25009551-25009551 on Build GRCh38
rs139054129 Chr.20: 25009317-25009317 on Build GRCh38
rs569921992 Chr.20: 25009240-25009240 on Build GRCh38
rs199734909 Chr.20: 25009299-25009299 on Build GRCh38
rs967181448 Chr.20: 25009181-25009181 on Build GRCh38
rs771200283 Chr.20: 25009297-25009297 on Build GRCh38
rs769379539 Chr.20: 25009272-25009272 on Build GRCh38
rs773433240 Chr.20: 25009291-25009291 on Build GRCh38
rs772006285 Chr.20: 25009481-25009481 on Build GRCh38
rs139694139 Chr.20: 25009597-25009597 on Build GRCh38
rs779759527 Chr.20: 25009273-25009273 on Build GRCh38
rs751734857 Chr.20: 25009354-25009354 on Build GRCh38
rs376502809 Chr.20: 25009306-25009306 on Build GRCh38
rs773023676 Chr.20: 25009422-25009422 on Build GRCh38
rs967274693 Chr.20: 25009586-25009586 on Build GRCh38
rs933076555 Chr.20: 25009275-25009275 on Build GRCh38
rs907207833 Chr.20: 25009278-25009278 on Build GRCh38
rs769708482 Chr.20: 25009392-25009392 on Build GRCh38
rs978064081 Chr.20: 25009507-25009507 on Build GRCh38
rs958036672 Chr.20: 25009460-25009460 on Build GRCh38
rs559350112 Chr.20: 25009413-25009413 on Build GRCh38
rs747784151 Chr.20: 25009414-25009414 on Build GRCh38
rs373840812 Chr.20: 25009330-25009330 on Build GRCh38
rs535322244 Chr.20: 25009251-25009251 on Build GRCh38
rs148489017 Chr.20: 25009215-25009215 on Build GRCh38

View Full Product Details