Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 25013874-25014374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSS1
Entrez Gene ID
84532
Gene Name
acyl-CoA synthetase short-chain family member 1
Gene Aliases
ACAS2L, ACECS1, AceCS2L
UniGene
Hs.529353
Gene Chromosome Location
Chr.20: 25006230-25058182 on Build GRCh38
Transcripts
NM_001252675.1 NM_001252676.1 NM_001252677.1 NM_032501.3 XM_006723659.1 XM_011529388.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3544851 c.1146C>T p.L382L Chr.20: 25014018-25014018 on Build GRCh38
2887290 c.1381G>A p.G461R Chr.20: 25014032-25014032 on Build GRCh38
5614760 c.1136C>A p.A379E Chr.20: 25014028-25014028 on Build GRCh38
4786628 c.1170C>G p.F390L Chr.20: 25013994-25013994 on Build GRCh38
3840604 c.1361C>T p.A454V Chr.20: 25014052-25014052 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767492073 Chr.20: 25013928-25013928 on Build GRCh38
rs201512408 Chr.20: 25013911-25013911 on Build GRCh38
rs149291639 Chr.20: 25013992-25013992 on Build GRCh38
rs544638695 Chr.20: 25014237-25014237 on Build GRCh38
rs369748334 Chr.20: 25013922-25013922 on Build GRCh38
rs949790532 Chr.20: 25014014-25014014 on Build GRCh38
rs200294586 Chr.20: 25014040-25014040 on Build GRCh38
rs779289465 Chr.20: 25014292-25014292 on Build GRCh38
rs182399293 Chr.20: 25013942-25013942 on Build GRCh38
rs776754978 Chr.20: 25014089-25014089 on Build GRCh38
rs376554358 Chr.20: 25014105-25014105 on Build GRCh38
rs766410829 Chr.20: 25014081-25014081 on Build GRCh38
rs772353096 Chr.20: 25014068-25014068 on Build GRCh38
rs553554587 Chr.20: 25014080-25014080 on Build GRCh38
rs758166369 Chr.20: 25014111-25014111 on Build GRCh38
rs373479533 Chr.20: 25013948-25013948 on Build GRCh38
rs1008881192 Chr.20: 25014234-25014234 on Build GRCh38
rs937018086 Chr.20: 25014350-25014350 on Build GRCh38
rs530425238 Chr.20: 25014286-25014286 on Build GRCh38
rs138608122 Chr.20: 25013984-25013984 on Build GRCh38
rs1010861293 Chr.20: 25014097-25014097 on Build GRCh38
rs183352912 Chr.20: 25014166-25014166 on Build GRCh38
rs776378158 Chr.20: 25013965-25013965 on Build GRCh38
rs540879771 Chr.20: 25014298-25014298 on Build GRCh38
rs766512466 Chr.20: 25014178-25014178 on Build GRCh38
rs374974276 Chr.20: 25014046-25014046 on Build GRCh38
rs773719841 Chr.20: 25014077-25014077 on Build GRCh38
rs760466942 Chr.20: 25014163-25014163 on Build GRCh38
rs144154135 Chr.20: 25013979-25013979 on Build GRCh38
rs375554533 Chr.20: 25014007-25014007 on Build GRCh38
rs758783320 Chr.20: 25014342-25014342 on Build GRCh38
rs763848096 Chr.20: 25014112-25014112 on Build GRCh38
rs552720099 Chr.20: 25014331-25014331 on Build GRCh38
rs371079525 Chr.20: 25014050-25014050 on Build GRCh38
rs766334344 Chr.20: 25014038-25014038 on Build GRCh38
rs192033932 Chr.20: 25014047-25014047 on Build GRCh38
rs538728681 Chr.20: 25013945-25013945 on Build GRCh38
rs187165890 Chr.20: 25013956-25013956 on Build GRCh38
rs759311286 Chr.20: 25013973-25013973 on Build GRCh38
rs371255511 Chr.20: 25014027-25014027 on Build GRCh38
rs952173571 Chr.20: 25014301-25014301 on Build GRCh38
rs777587427 Chr.20: 25013950-25013950 on Build GRCh38
rs532635170 Chr.20: 25014308-25014308 on Build GRCh38
rs780837257 Chr.20: 25014125-25014125 on Build GRCh38
rs202004508 Chr.20: 25014028-25014028 on Build GRCh38
rs746946586 Chr.20: 25014039-25014039 on Build GRCh38
rs753812015 Chr.20: 25014214-25014214 on Build GRCh38
rs545969244 Chr.20: 25014133-25014133 on Build GRCh38
rs755065708 Chr.20: 25014285-25014285 on Build GRCh38
rs751511903 Chr.20: 25014118-25014118 on Build GRCh38
rs750572630 Chr.20: 25013929-25013929 on Build GRCh38
rs114969892 Chr.20: 25014300-25014300 on Build GRCh38
rs753065403 Chr.20: 25013985-25013985 on Build GRCh38
rs757332481 Chr.20: 25014120-25014120 on Build GRCh38
rs775016558 Chr.20: 25014053-25014053 on Build GRCh38
rs756161241 Chr.20: 25013930-25013930 on Build GRCh38
rs780183444 Chr.20: 25013936-25013936 on Build GRCh38
rs79644787 Chr.20: 25014180-25014180 on Build GRCh38
rs949852426 Chr.20: 25014252-25014252 on Build GRCh38
rs770600238 Chr.20: 25013958-25013958 on Build GRCh38
rs561230366 Chr.20: 25014244-25014244 on Build GRCh38
rs751988730 Chr.20: 25013917-25013917 on Build GRCh38
rs138245840 Chr.20: 25013975-25013975 on Build GRCh38
rs918391751 Chr.20: 25014251-25014251 on Build GRCh38
rs972979651 Chr.20: 25014232-25014232 on Build GRCh38
rs112683865 Chr.20: 25013978-25013978 on Build GRCh38
rs774820049 Chr.20: 25013972-25013972 on Build GRCh38
rs900530994 Chr.20: 25014247-25014247 on Build GRCh38
rs909838420 Chr.20: 25014189-25014189 on Build GRCh38
rs932586269 Chr.20: 25013964-25013964 on Build GRCh38
rs953403264 Chr.20: 25014179-25014179 on Build GRCh38
rs754032909 Chr.20: 25014006-25014006 on Build GRCh38
rs374524367 Chr.20: 25014054-25014054 on Build GRCh38
rs1042295893 Chr.20: 25014169-25014169 on Build GRCh38
rs768008909 Chr.20: 25013982-25013982 on Build GRCh38
rs766469777 Chr.20: 25013999-25013999 on Build GRCh38
rs1020815670 Chr.20: 25013941-25013941 on Build GRCh38
rs148688525 Chr.20: 25014126-25014126 on Build GRCh38
rs750401692 Chr.20: 25014106-25014106 on Build GRCh38
rs778565258 Chr.20: 25014011-25014011 on Build GRCh38
rs879468846 Chr.20: 25013996-25013996 on Build GRCh38

View Full Product Details