Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 25302097-25302597 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD12
Entrez Gene ID
26090
Gene Name
abhydrolase domain containing 12
Gene Aliases
ABHD12A, BEM46L2, C20orf22, PHARC, dJ965G21.2
UniGene
Hs.441550
Gene Chromosome Location
Chr.20: 25294743-25390982 on Build GRCh38
Transcripts
NM_001042472.2 NM_015600.4 XM_011529214.2 XM_017027796.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1025345 c.1144C>T p.P382S Chr.20: 25302232-25302232 on Build GRCh38
1592507 c.1144C>T p.P382S Chr.20: 25302232-25302232 on Build GRCh38
1025346 c.1100A>G p.D367G Chr.20: 25302276-25302276 on Build GRCh38
3933814 c.1044C>T p.A348A Chr.20: 25302332-25302332 on Build GRCh38
1025347 c.1030-1G>T p.? Chr.20: 25302347-25302347 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746748 Chr.20: 25302331-25302331 on Build GRCh38
rs939197468 Chr.20: 25302560-25302560 on Build GRCh38
rs371663818 Chr.20: 25302185-25302185 on Build GRCh38
rs772788826 Chr.20: 25302463-25302463 on Build GRCh38
rs537316496 Chr.20: 25302502-25302502 on Build GRCh38
rs781754739 Chr.20: 25302352-25302353 on Build GRCh38
rs974325146 Chr.20: 25302443-25302443 on Build GRCh38
rs758755514 Chr.20: 25302373-25302373 on Build GRCh38
rs986928377 Chr.20: 25302419-25302419 on Build GRCh38
rs200023360 Chr.20: 25302169-25302169 on Build GRCh38
rs776404400 Chr.20: 25302321-25302321 on Build GRCh38
rs376953929 Chr.20: 25302312-25302312 on Build GRCh38
rs866494149 Chr.20: 25302128-25302128 on Build GRCh38
rs529587366 Chr.20: 25302505-25302505 on Build GRCh38
rs949742689 Chr.20: 25302544-25302544 on Build GRCh38
rs1003468289 Chr.20: 25302221-25302221 on Build GRCh38
rs1024211064 Chr.20: 25302135-25302135 on Build GRCh38
rs587777602 Chr.20: 25302260-25302260 on Build GRCh38
rs1034960218 Chr.20: 25302354-25302354 on Build GRCh38
rs147060658 Chr.20: 25302238-25302238 on Build GRCh38
rs146028040 Chr.20: 25302263-25302263 on Build GRCh38
rs747382300 Chr.20: 25302277-25302277 on Build GRCh38
rs765988372 Chr.20: 25302229-25302229 on Build GRCh38
rs925934756 Chr.20: 25302181-25302181 on Build GRCh38
rs890128966 Chr.20: 25302162-25302162 on Build GRCh38
rs745930769 Chr.20: 25302366-25302397 on Build GRCh38
rs886315445 Chr.20: 25302208-25302208 on Build GRCh38
rs781689566 Chr.20: 25302183-25302183 on Build GRCh38
rs756448048 Chr.20: 25302341-25302341 on Build GRCh38
rs756681155 Chr.20: 25302265-25302265 on Build GRCh38
rs748330820 Chr.20: 25302273-25302273 on Build GRCh38
rs770597033 Chr.20: 25302387-25302387 on Build GRCh38
rs766152337 Chr.20: 25302500-25302500 on Build GRCh38
rs766949743 Chr.20: 25302346-25302346 on Build GRCh38
rs116792881 Chr.20: 25302521-25302521 on Build GRCh38
rs773891664 Chr.20: 25302333-25302333 on Build GRCh38
rs548270757 Chr.20: 25302486-25302486 on Build GRCh38
rs748383973 Chr.20: 25302367-25302367 on Build GRCh38
rs765033798 Chr.20: 25302561-25302561 on Build GRCh38
rs757590387 Chr.20: 25302172-25302172 on Build GRCh38
rs200536497 Chr.20: 25302313-25302313 on Build GRCh38
rs557573795 Chr.20: 25302206-25302206 on Build GRCh38
rs757670984 Chr.20: 25302301-25302301 on Build GRCh38
rs1013369297 Chr.20: 25302405-25302405 on Build GRCh38
rs149514933 Chr.20: 25302293-25302293 on Build GRCh38
rs774414174 Chr.20: 25302216-25302216 on Build GRCh38
rs746844858 Chr.20: 25302384-25302384 on Build GRCh38
rs1000245834 Chr.20: 25302441-25302441 on Build GRCh38
rs267606624 Chr.20: 25302322-25302322 on Build GRCh38
rs529522527 Chr.20: 25302132-25302132 on Build GRCh38
rs964407117 Chr.20: 25302487-25302487 on Build GRCh38
rs181420034 Chr.20: 25302474-25302474 on Build GRCh38
rs368948114 Chr.20: 25302187-25302187 on Build GRCh38
rs542939526 Chr.20: 25302268-25302268 on Build GRCh38
rs898798041 Chr.20: 25302301-25302301 on Build GRCh38
rs371418239 Chr.20: 25302332-25302332 on Build GRCh38
rs987669200 Chr.20: 25302470-25302470 on Build GRCh38
rs776269106 Chr.20: 25302231-25302231 on Build GRCh38
rs995113615 Chr.20: 25302198-25302198 on Build GRCh38
rs778098121 Chr.20: 25302226-25302226 on Build GRCh38
rs761264673 Chr.20: 25302375-25302375 on Build GRCh38
rs369745821 Chr.20: 25302327-25302327 on Build GRCh38
rs778798173 Chr.20: 25302361-25302361 on Build GRCh38
rs767189683 Chr.20: 25302452-25302452 on Build GRCh38
rs942963737 Chr.20: 25302428-25302428 on Build GRCh38
rs895806378 Chr.20: 25302365-25302365 on Build GRCh38
rs776655670 Chr.20: 25302395-25302395 on Build GRCh38
rs763733941 Chr.20: 25302334-25302334 on Build GRCh38
rs757643586 Chr.20: 25302254-25302254 on Build GRCh38
rs750935234 Chr.20: 25302264-25302264 on Build GRCh38
rs1003439143 Chr.20: 25302348-25302348 on Build GRCh38
rs530146493 Chr.20: 25302414-25302414 on Build GRCh38
rs1039217171 Chr.20: 25302165-25302165 on Build GRCh38
rs752001929 Chr.20: 25302187-25302187 on Build GRCh38
rs779225002 Chr.20: 25302272-25302272 on Build GRCh38
rs761738607 Chr.20: 25302217-25302217 on Build GRCh38
rs553242935 Chr.20: 25302297-25302297 on Build GRCh38
rs570352851 Chr.20: 25302491-25302491 on Build GRCh38
rs10966 Chr.20: 25302308-25302308 on Build GRCh38
rs543440298 Chr.20: 25302450-25302450 on Build GRCh38
rs749432855 Chr.20: 25302266-25302266 on Build GRCh38
rs142709197 Chr.20: 25302235-25302235 on Build GRCh38
rs911350780 Chr.20: 25302422-25302422 on Build GRCh38
rs746328599 Chr.20: 25302184-25302184 on Build GRCh38
rs755106705 Chr.20: 25302358-25302358 on Build GRCh38
rs969169126 Chr.20: 25302438-25302438 on Build GRCh38
rs752254456 Chr.20: 25302247-25302247 on Build GRCh38
rs771951512 Chr.20: 25302220-25302220 on Build GRCh38
rs991573752 Chr.20: 25302232-25302232 on Build GRCh38
rs549214257 Chr.20: 25302149-25302149 on Build GRCh38
rs920279895 Chr.20: 25302499-25302499 on Build GRCh38
rs533451545 Chr.20: 25302449-25302449 on Build GRCh38
rs772002701 Chr.20: 25302288-25302288 on Build GRCh38
rs188888939 Chr.20: 25302335-25302335 on Build GRCh38
rs139135254 Chr.20: 25302228-25302228 on Build GRCh38
rs771364543 Chr.20: 25302289-25302289 on Build GRCh38
rs4358194 Chr.20: 25302117-25302117 on Build GRCh38
rs373615018 Chr.20: 25302224-25302224 on Build GRCh38
rs62212154 Chr.20: 25302164-25302164 on Build GRCh38
rs754258279 Chr.20: 25302353-25302353 on Build GRCh38
rs752849584 Chr.20: 25302566-25302566 on Build GRCh38
rs759121223 Chr.20: 25302329-25302329 on Build GRCh38
rs749323860 Chr.20: 25302188-25302188 on Build GRCh38

View Full Product Details