Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CWC15
Entrez Gene ID
51503
Gene Name
CWC15 spliceosome-associated protein
Gene Aliases
AD002, C11orf5, Cwf15, HSPC148, ORF5
Chromosome Location
Chr.11: 94962622 - 94973612 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016403.3 NP_057487.2 6 - 7 686 104
XM_011542868.2 XP_011541170.1 6 - 7 821 104
XM_017017916.1 XP_016873405.1 6 - 7 874 104
GenBank mRNA AF110775.1 - 6 - 7 626 104
AF161497.1 - 6 - 7 626 104
AJ250393.1 - 6 - 7 643 104
AK225956.1 - 6 - 7 645 104
AK314900.1 - 6 - 7 630 104
BC040946.1 - 6 - 7 638 104
BM971024.1 - 1 - 2 104
BU676427.1 - 1 - 2 104

View Full Product Details