Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IGF2-AS
Entrez Gene ID
51214
Gene Name
IGF2 antisense RNA
Gene Aliases
IGF2-AS1, IGF2AS, PEG8
Chromosome Location
Chr.11: 2140512 - 2148666 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_028043.2 1 - 2 436 107
GenBank mRNA AL521915.3 - 1 - 2 430 107

View Full Product Details