Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33066740-33067246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFB3
Entrez Gene ID
359710
Gene Name
BPI fold containing family B member 3
Gene Aliases
C20orf185, LPLUNC3, RYA3
UniGene
Hs.472442
Gene Chromosome Location
Chr.20: 33055331-33073637 on Build GRCh38
Transcripts
NM_182658.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4950952 c.941C>A p.P314Q Chr.20: 33066840-33066840 on Build GRCh38
212042 c.976G>A p.E326K Chr.20: 33066875-33066875 on Build GRCh38
3972515 c.977A>G p.E326G Chr.20: 33066876-33066876 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs763601079 Chr.20: 33066830-33066830 on Build GRCh38
rs1029514206 Chr.20: 33067129-33067129 on Build GRCh38
rs766690906 Chr.20: 33066835-33066835 on Build GRCh38
rs570129503 Chr.20: 33067211-33067211 on Build GRCh38
rs967055183 Chr.20: 33066840-33066840 on Build GRCh38
rs560921400 Chr.20: 33067054-33067054 on Build GRCh38
rs772259493 Chr.20: 33066804-33066804 on Build GRCh38
rs150087753 Chr.20: 33066832-33066832 on Build GRCh38
rs1006647581 Chr.20: 33067155-33067155 on Build GRCh38
rs949396233 Chr.20: 33067002-33067002 on Build GRCh38
rs756638010 Chr.20: 33066833-33066833 on Build GRCh38
rs541333265 Chr.20: 33067163-33067163 on Build GRCh38
rs55668794 Chr.20: 33066893-33066893 on Build GRCh38
rs778018376 Chr.20: 33066880-33066880 on Build GRCh38
rs375629521 Chr.20: 33066890-33066890 on Build GRCh38
rs542846523 Chr.20: 33067046-33067046 on Build GRCh38
rs1056172862 Chr.20: 33067089-33067089 on Build GRCh38
rs937461865 Chr.20: 33067078-33067078 on Build GRCh38
rs746953454 Chr.20: 33066811-33066811 on Build GRCh38
rs926723441 Chr.20: 33067016-33067016 on Build GRCh38
rs202121541 Chr.20: 33066849-33066849 on Build GRCh38
rs764778155 Chr.20: 33066817-33066817 on Build GRCh38
rs1018319014 Chr.20: 33067160-33067160 on Build GRCh38
rs894082960 Chr.20: 33067057-33067057 on Build GRCh38
rs547687839 Chr.20: 33067145-33067145 on Build GRCh38
rs753958044 Chr.20: 33067032-33067032 on Build GRCh38
rs138544042 Chr.20: 33066844-33066844 on Build GRCh38
rs1043616020 Chr.20: 33067109-33067109 on Build GRCh38
rs996695846 Chr.20: 33067157-33067157 on Build GRCh38
rs762681428 Chr.20: 33066929-33066929 on Build GRCh38
rs554058602 Chr.20: 33067003-33067003 on Build GRCh38
rs772342821 Chr.20: 33066873-33066874 on Build GRCh38
rs531859224 Chr.20: 33067066-33067066 on Build GRCh38
rs145623298 Chr.20: 33067096-33067096 on Build GRCh38
rs138216372 Chr.20: 33067201-33067201 on Build GRCh38
rs1011570540 Chr.20: 33067100-33067100 on Build GRCh38
rs762367698 Chr.20: 33066827-33066827 on Build GRCh38
rs369627899 Chr.20: 33066815-33066815 on Build GRCh38
rs187134945 Chr.20: 33067167-33067167 on Build GRCh38
rs763684455 Chr.20: 33067128-33067128 on Build GRCh38
rs376864545 Chr.20: 33066812-33066812 on Build GRCh38
rs749462944 Chr.20: 33066774-33066774 on Build GRCh38
rs532343653 Chr.20: 33067141-33067141 on Build GRCh38
rs902883165 Chr.20: 33067114-33067114 on Build GRCh38
rs548812143 Chr.20: 33067209-33067209 on Build GRCh38
rs746898310 Chr.20: 33066886-33066886 on Build GRCh38
rs372706723 Chr.20: 33066904-33066904 on Build GRCh38
rs755124114 Chr.20: 33066783-33066783 on Build GRCh38
rs908567185 Chr.20: 33066986-33066986 on Build GRCh38
rs918884645 Chr.20: 33066930-33066930 on Build GRCh38
rs113470611 Chr.20: 33066997-33066997 on Build GRCh38
rs113141530 Chr.20: 33066907-33066907 on Build GRCh38
rs779243397 Chr.20: 33066785-33066785 on Build GRCh38
rs372453694 Chr.20: 33066860-33066860 on Build GRCh38
rs754179740 Chr.20: 33066836-33066836 on Build GRCh38
rs533770373 Chr.20: 33067159-33067160 on Build GRCh38
rs902467260 Chr.20: 33067120-33067120 on Build GRCh38
rs1009800520 Chr.20: 33067088-33067088 on Build GRCh38
rs985505478 Chr.20: 33066879-33066879 on Build GRCh38
rs1034378761 Chr.20: 33066778-33066778 on Build GRCh38
rs748162557 Chr.20: 33066791-33066791 on Build GRCh38
rs955171811 Chr.20: 33066857-33066857 on Build GRCh38
rs543535251 Chr.20: 33067075-33067075 on Build GRCh38
rs773215390 Chr.20: 33066807-33066807 on Build GRCh38
rs775273983 Chr.20: 33066819-33066819 on Build GRCh38
rs1007891956 Chr.20: 33067125-33067125 on Build GRCh38
rs182099730 Chr.20: 33067036-33067036 on Build GRCh38
rs770964375 Chr.20: 33066887-33066887 on Build GRCh38
rs370006680 Chr.20: 33066841-33066841 on Build GRCh38
rs760779143 Chr.20: 33067014-33067014 on Build GRCh38
rs373286640 Chr.20: 33066816-33066816 on Build GRCh38
rs766778345 Chr.20: 33067026-33067026 on Build GRCh38

View Full Product Details